All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H4O4
CAS Number:
Molecular Weight:
164.11
Beilstein:
135783
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
98%
form
solid
mp
199-202 °C (lit.)
SMILES string
Oc1cccc2C(=O)OC(=O)c12
InChI
1S/C8H4O4/c9-5-3-1-2-4-6(5)8(11)12-7(4)10/h1-3,9H
InChI key
CCTOEAMRIIXGDJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lin Li et al.
Journal of acquired immune deficiency syndromes (1999), 56(5), 384-392 (2011-01-18)
The development of a safe, effective, and affordable microbicide to prevent the sexual transmission of HIV combination is urgently needed. Our previous studies demonstrated that 3-hydroxyphthalic anhydride-modified chicken ovalbumin (HP-OVA) exhibited potent antiviral activity against a broad spectrum of HIV
Lin Li et al.
The Journal of antimicrobial chemotherapy, 68(3), 573-576 (2012-12-12)
We recently demonstrated that both 3-hydroxyphthalic anhydride (HP)- and maleic anhydride-modified chicken ovalbumin (OVA) could effectively inhibit HIV-1 infection. But because OVA may cause allergy in some human subjects, here we replaced OVA with human serum albumin (HSA) in designing
Youming Shen et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 185, 371-375 (2017-06-12)
A new turn-on phthalimide fluorescent probe has designed and synthesized for sensing cysteine (Cys) based on excited state intramolecular proton transfer (ESIPT) process. It is consisted of a 3-hydroxyphthalimide derivative moiety as the fluorophore and an acrylic ester group as
Anna Oevermann et al.
Antiviral research, 59(1), 23-33 (2003-07-02)
Chemical modification of the proteins bovine serum albumin, alpha-lactalbumin, beta-lactoglobulin and chicken lysozyme by 3-hydroxyphthalic anhydride (3-HP) yielded compounds which exerted antiviral activity in vitro as compared with the native unmodified proteins. Of the three enveloped viruses tested, human herpes
Li-li He et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 31(7), 1175-1178 (2011-07-19)
To investigate the antiviral activity of 3-hydroxyphthalic anhydride-modified ovalbumin (HP-OVA) against herpes simplex virus 2 (HSV-2) in vitro. By chemical modification, ovalbumin (OVA) was treated with 3-hydroxyphthalic anhydride (HP) to prepare HP-OVA. The anti-HSV-2 activity against HSV-2 333 virus in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service