Skip to Content
Merck
All Photos(3)

Key Documents

307823

Sigma-Aldrich

Sodium hydrogen sulfate

technical grade

Synonym(s):

Sodium bisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHSO4
CAS Number:
Molecular Weight:
120.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
technical grade
form:
crystalline
Pricing and availability is not currently available.

grade

technical grade

Assay

≥90.0% NaOH basis (titration)

form

crystalline

SMILES string

[Na+].OS([O-])(=O)=O

InChI

1S/Na.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1

InChI key

WBHQBSYUUJJSRZ-UHFFFAOYSA-M

General description

Sodium hydrogen sulfate, also known as sodium bisulfate, is commonly used as a pH modifier, acidulant, and chemical reagent in various industrial and laboratory applications.

Application

Sodium hydrogen sulfate can be used as:      
  • A source of sulfur in the synthesis of the Ni-doped Cu2ZnSnS4 (CZTS) material, which find its application in solar energy conversion technologies.
  • As a pH regulator to maintain acidic conditions in various processes.   
  • A acidic catalyst to facilitate the esterification process between oleyl alcohol and oleic acid.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Sato et al.
Veterinary and comparative oncology, 16(3), 409-415 (2018-03-13)
Death-associated protein kinase (DAPK) is a serine/threonine kinase and a tumour suppressor gene. Diffuse large B-cell lymphomas with inactivated DAPK through hypermethylation of a CpG island is known to result in a biologically aggressive phenotype in humans. This retrospective study
Shanshan Li et al.
Life sciences, 231, 116576-116576 (2019-06-19)
Studies suggest that cardiovascular function in offspring can be epigenetically programmed by environmental changes during pregnancy. CaV1.2 channel plays a major role in the regulation of the vascular tone. This study investigated the effects and underlying mechanisms of exercise during
Shima Borzouie et al.
Animals : an open access journal from MDPI, 10(9) (2020-09-10)
This study was planned to investigate the effects of seaweed supplementation, genetic strain, heat stress and their interactions on laying hen performances, blood chemistry and hematology. In a short-term trial, laying hens of the two genetic lines Lohman LSL-Lite (White)
Elisabet Ihse et al.
Scientific reports, 7(1), 9008-9008 (2017-08-23)
Amyloid aggregates found in the brain of patients with neurodegenerative diseases, including Alzheimer's and Parkinson's disease, are thought to spread to increasingly larger areas of the brain through a prion-like seeding mechanism. Not much is known about which cell surface
Fabrication of self-ordered porous alumina via anodizing in sulfate solutions
Kondo R, et al.
Materials Letters, 183, 285-289 (2016)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service