Skip to Content
Merck
All Photos(1)

Documents

301590

Sigma-Aldrich

Tetrabutylammonium tribromide

98%

Synonym(s):

TBABr3, Tetrabutylammonium bromide perbromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4NBr3
CAS Number:
Molecular Weight:
482.18
Beilstein:
3746114
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

71-76 °C (lit.)

SMILES string

Br[Br-]Br.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.Br3/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-3-2/h5-16H2,1-4H3;/q+1;-1

InChI key

XXSLZJZUSYNITM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrabutylammonium tribromide can be used:
  • As a catalyst in the preparation of O-isopropylidene derivatives of free sugars.
  • As a reagent in methanol for the cleavage of tert-butyldimethylsilyl (TBDMS) ethers chemoselectively.
  • As a catalyst in the synthesis of bis-indolymethanes by electrophilic substitution of indoles with different aldehydes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrabutylammonium tribromide (TBATB)- MeOH: An efficient chemoselective reagent for the cleavage of tert-butyldimethylsilyl (TBDMS) ethers
Gopinath R and Patel BK
Organic Letters, 2(26), 4177-4180 (2000)
Mild and Efficient Synthesis of bis-Indolylmethanes Catalyzed by Tetrabutylammonium Tribromide
Lin X, et al.
Synthetic Communications, 36(21), 3153-3160 (2006)
Tetrabutylammonium tribromide (TBATB): a mild and efficient catalyst for O-isopropylidenation of carbohydrates
Khan AT, et al.
Carbohydrate Research, 346(5), 673-677 (2011)
Shigeo Hayashi et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 846-867 (2014-02-13)
Because of the pivotal role of cyclooxygenase (COX) in the inflammatory processes, non-steroidal anti-inflammatory drugs (NSAIDs) that suppress COX activities have been used clinically for the treatment of inflammatory diseases/syndromes; however, traditional NSAIDs exhibit serious side-effects such as gastrointestinal damage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service