Skip to Content
Merck
All Photos(2)

Documents

300349

Sigma-Aldrich

(2S,3S)-(+)-2,3-Butanediol

97%

Synonym(s):

(+)-2,3-Butanediol, (2S,3S)-2,3-Butanediol, (S,S)-2,3-Butanediol, Dextro-2,3-Butanediol, L-(+)-Butane-2,3-diol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(OH)CH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1718899
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

optical activity

[α]20/D +13°, neat

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.433 (lit.)

bp

179-182 °C (lit.)

density

0.987 g/mL at 25 °C (lit.)

SMILES string

C[C@H](O)[C@H](C)O

InChI

1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3/t3-,4-/m0/s1

InChI key

OWBTYPJTUOEWEK-IMJSIDKUSA-N

General description

(2S,3S)-2,3-Butanediol ((2S,3S)-2,3-BD) is a chiral compound that can be used as a building block in organic synthesis for the preparation of bioactive molecules.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biocatalytic production of (2S, 3S)-2, 3-butanediol from diacetyl using whole cells of engineered Escherichia coli
Li L, et al.
Bioresource Technology, 115, 111-116 (2012)
Sang Heum Shin et al.
Journal of bacteriology, 194(10), 2736-2737 (2012-04-27)
Here we report the full genome sequence of Klebsiella pneumoniae KCTC 2242,consisting of a 5.26-Mb chromosome (57.6% GC%; 5,035 genes [4,923 encoding known proteins, 112 RNA genes]) and a 202-kb plasmid (50.2% GC%; 229 genes [229 encoding known proteins]).
Yuansheng Xue et al.
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 27(12), 1742-1748 (2012-04-18)
The production of 2, 3-butanediol and succinic acid by a moderate halophile under anaerobic condition was investigated. This halophile, termed Salinivibrio YS, was isolated from the solid samples collected from Aydingkol Lake. Based on the single factor experiment, the parameters
Mingshou Lu et al.
Journal of microbiology and biotechnology, 22(5), 659-667 (2012-05-09)
2,3-Butanediol (2,3-BDO) is an organic compound with a wide range of industrial applications. Although Escherichia coli is often used for the production of organic compounds, the wild-type E. coli does not contain two essential genes in the 2,3-BDO biosynthesis pathway
Jinshan Li et al.
Applied microbiology and biotechnology, 97(2), 585-597 (2012-08-07)
Paenibacillus polymyxa can produce the (R,R)-stereoisomer of 2,3-butanediol (2,3-BDL) which is industrially very useful. Two important factors affecting (R,R)-BDL production by P. polymyxa ATCC 12321, medium composition, and addition of acetic acid to the culture were investigated in this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service