Skip to Content
Merck
All Photos(3)

Key Documents

294438

Sigma-Aldrich

4-Nitrobenzenediazonium tetrafluoroborate

97%

Synonym(s):

Azoic Diazo No. 37, Fast Red GG salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4N3O2 · BF4
CAS Number:
Molecular Weight:
236.92
Beilstein:
3599869
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

form

powder

composition

Carbon 29.3-31.5%,Nitrogen 17.1-18.4%

technique(s)

titration: suitable

color

light brown

mp

144-148 °C (dec.)

solubility

acetonitrile: 50 mg/mL

density

1.66 g/cm3 at 25 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.[O-][N+](=O)c1ccc(cc1)[N+]#N

InChI

1S/C6H4N3O2.BF4/c7-8-5-1-3-6(4-2-5)9(10)11;2-1(3,4)5/h1-4H;/q+1;-1

InChI key

AFULQCYCQAHYIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitrobenzenediazonium tetrafluoroborate, also known as Fast Red GG is a Diazonium salt/diazo dye. It is commonly used for the histochemical detection of bilirubin and indolamines.

Application

4-Nitrobenzenediazonium tetrafluoroborate has been used in ellipsometry. It has also been used for surface modification of carbon nanotubes (CNTs).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transparent conducting films made of different carbon nanotubes, processed carbon nanotubes, and graphene nanoribbons
Anson-Casaos A, et al.
Chemical Engineering Science, 138(1), 566-574 (2015)
Y Harano et al.
Clinica chimica acta; international journal of clinical chemistry, 134(3), 327-336 (1983-11-15)
A highly sensitive and simplified method for the differential determination of serum ketone bodies has been developed. Serum was deproteinized with perchloric acid, and acetoacetate contained in the supernate was reacted with newly synthesized p-nitrobenzene diazonium fluoroborate at 37 degrees
E N Petrova et al.
Biokhimiia (Moscow, Russia), 51(10), 1732-1740 (1986-10-01)
It was found that at pH 5.2 and 40-fold excess of p-nitrophenyldiazonium chloride the inhibitor incorporation into the porcine pepsin molecule involves 1.9 residues, one residue being bound to tyrosine 189. Besides, tyrosines 44, 113, 154 and 174 enter the
Electrochemical grafting on SOI substrates using aryl diazonium salts
Le Floch F, et al.
Journal of Electroanalytical Chemistry, 660(1), 127-132 (2011)
Zari Tehrani et al.
Nanomaterials (Basel, Switzerland), 11(2) (2021-02-11)
Electrochemical biosensors using carbon-based electrodes are being widely developed for the detection of a range of different diseases. Since their sensitivity depends on the surface coverage of bioreceptor moieties, it necessarily depends on the surface coverage of amine precursors. Electrochemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service