Skip to Content
Merck
All Photos(1)

Key Documents

289205

Supelco

Lanthanum(III) oxide

suitable for AAS, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La2O3
CAS Number:
Molecular Weight:
325.81
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Agency

meets analytical specifications of BP
meets analytical specifications of E252
meets analytical specifications of Ph. Eur.
meets analytical specifications of USP

reg. compliance

meets analytical specifications of FCC

Assay

≥99.9% trace metals basis
≥99.9%

form

powder

reaction suitability

reagent type: catalyst
core: lanthanum

technique(s)

AAS: suitable

density

6.51 g/mL at 25 °C (lit.)

SMILES string

O=[La]O[La]=O

InChI

1S/2La.3O

InChI key

KTUFCUMIWABKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lanthanum (III) oxide is an alkaline earth oxide, and is a diamagnetic, isostructural analog of neodymium oxide. It is also an efficient recyclable catalyst, and can be used in coupling various aryl halides with aromatic/alkyl thiols.

Application

Lanthanum (III) oxide was used as an analytical standard in inductively coupled plasma atomic emission spectrometry (ICP-AES) for chemical characterization of ancient pottery.
Precursor to LAMOX fast ion conductors and superconductors.
Suitable for use as atomic absorption standard.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted versus conventional decomposition procedures applied to a ceramic potsherd standard reference material by inductively coupled plasma atomic emission spectrometry.
Papadopoulou DN
Analytica Chimica Acta, 505 (1), 173-181 (2004)
Aldol addition of acetone, catalyzed by solid base catalysts: magnesium oxide, calcium oxide, strontium oxide, barium oxide, lanthanum (III) oxide and zirconium oxide.
Zhang G
Applied Catalysis, 36, 189-197 (1988)
THERMOPHYSICAL PROPERTIES OF THE LANTHANIDE OXIDES. I. HEAT CAPACITIES, THERMODYNAMIC PROPERTIES, AND SOME ENERGY LEVELS OF LANTHANUM (III) AND NEODYMIUM (III) OXIDES FROM 5 TO 350? K.
The Journal of Physical Chemistry, 67 (2), 339-345 (1963)
Lanthanum (III) oxide as a recyclable catalyst for the synthesis of diaryl sulfides and diaryl selenides.
The Journal of Organic Chemistry, 34, 5902-5905 (2009)
Anita J Vincent-Johnson et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(3), 035901-035901 (2012-12-12)
Upon excitation in thin oxide films by infrared radiation, radiative polaritons are formed with complex angular frequency ω, according to the theory of Kliewer and Fuchs (1966 Phys. Rev. 150 573). We show that radiative polaritons leak radiation with frequency

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service