287717
Borane-ammonia complex
technical grade, 90%
Synonym(s):
Amminetrihydroboron, Borazane
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
H3N · BH3
CAS Number:
Molecular Weight:
30.87
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Quality Level
Assay
90%
form
solid
reaction suitability
reagent type: reductant
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
SMILES string
B.N
InChI
1S/BH3.H3N/h2*1H3
InChI key
WZMUUWMLOCZETI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Ammonia borane, also known as borazane has high hydrogen content (about 20% mass). Thermal decomposition of borazane takes place in the temperature range of 350-410K accompanied by hydrogen release and heat evolution. It may find use as a hydrogen source in fuel cells in the future.
Borane-ammonia complex is a novel material, where ammonia is tightly bound to a ligand through all four of its atoms. It is a donor-acceptor complex that is formed by electron donation from a Lewis base (NH3) to a Lewis acid (BH3). It shows a B-N bond distance of 1.564 Å.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Application
Borazane acts as a mild, efficient, and stereoselective reducing agent for aldehydes and ketones in protic or nonprotic media.It can be used as a storage medium for release of hydrogenBorazane is popularly used as an ammonia borane (AB) source for the formation of hexagonal boron nitride monolayers which are used as a substrate for graphene growth.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Weng, B.; et al.
International Journal of Hydrogen Energy, 36, 10870-10870 (2011)
William C Ewing et al.
Journal of the American Chemical Society, 133(42), 17093-17099 (2011-10-04)
Studies of the activating effect of Verkade's base, 2,8,9-triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane (VB), on the rate and extent of H(2) release from ammonia borane (AB) have led to the syntheses and structural characterizations of three anionic aminoborane chain-growth products that provide direct support
X. Yang, et al.,
Tetrahedron, 67, 7121-7127 (2011)
Thermal decomposition of B-N-H compounds investigated by using combined thermoanalytical methods.
Baitalow F, et al.
Thermochimica Acta, 391(1), 159-168 (2002)
Gang Hee Han et al.
ACS nano, 7(11), 10129-10138 (2013-11-05)
Graphene-boron nitride monolayer heterostructures contain adjacent electrically active and insulating regions in a continuous, single-atom thick layer. To date structures were grown at low pressure, resulting in irregular shapes and edge direction, so studies of the graphene-boron nitride interface were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service