Skip to Content
Merck
All Photos(2)

Documents

287644

Sigma-Aldrich

(1R,2R)-(−)-Pseudoephedrine

98%

Synonym(s):

α-(1-Methylaminoethyl)benzyl alcohol, (−)-ψ-Ephedrine, (−)-Pseudoephedrine, (1R,2R)-(−)-2-(Methylamino)-1-phenylpropanol, l-Pseudoephedrine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH
CAS Number:
Molecular Weight:
165.23
Beilstein:
2803417
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

optical activity

[α]20/D −51°, c = 0.6 in ethanol

mp

118-120 °C (lit.)

SMILES string

CN[C@H](C)[C@H](O)c1ccccc1

InChI

1S/C10H15NO/c1-8(11-2)10(12)9-6-4-3-5-7-9/h3-8,10-12H,1-2H3/t8-,10+/m1/s1

InChI key

KWGRBVOPPLSCSI-SCZZXKLOSA-N

General description

(1R,2R)-(-)-Pseudoephedrine is an enantiomer of ephedrine mainly used as a chiral auxiliary for asymmetric synthesis.

Application

(1R,2R)-(-)-Pseudoephedrine may be used in the preparation of chiral relay ligands, which can catalyze the addition of diethylzinc to aldehydes leading to the corresponding secondary alcohol. The immobilization of pseudoephedrine on Merrifield resin forms a chiral linker, which may be used for the asymmetric alkylation of amides via solid-phase approach.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of a pseudoephedrine linker for asymmetric alkylations on solid phase.
Hutchison PC, et al.
Organic Letters, 4(26), 4583-4585 (2002)
Enantioselective diethylzinc additions to aldehydes catalyzed by chiral relay ligands.
Sibi MP and Levi M. Stanley.
Tetrahedron Asymmetry, 15(21), 3353-3356 (2004)
Pseudoephedrine as a practical chiral auxiliary for the synthesis of highly enantiomerically enriched carboxylic acids, alcohols, aldehydes, and ketones.
Myers AG, et al.
Journal of the American Chemical Society, 119(28), 6496-6511 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service