Skip to Content
Merck
All Photos(1)

Key Documents

285005

Sigma-Aldrich

D-Allose

98%

Synonym(s):

β-D-Allopyranose, β-D-Allose, All

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724623
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

powder

optical activity

[α]19/D +15°, c = 3.8 in H2O

mp

148-150 °C (lit.)

SMILES string

OC[C@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4-,5-,6-/m1/s1

InChI key

WQZGKKKJIJFFOK-QZABAPFNSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jianjun Xie et al.
International journal of oncology, 50(3), 863-872 (2017-02-16)
Osteosarcoma (OS) has a high degree of chromosomal instability and total copy number (CN) changes. We examined 58 human OS samples including 40 primary tumors, 11 explants, and 7 cell lines using single nucleotide polymorphism (SNP) arrays, and revealed that
Wilmer H Perera et al.
Molecules (Basel, Switzerland), 26(4) (2021-03-07)
Diabetes mellitus is a chronic disease and one of the fastest-growing health challenges of the last decades. Studies have shown that chronic low-grade inflammation and activation of the innate immune system are intimately involved in type 2 diabetes pathogenesis. Momordica
Stefano Motta et al.
Journal of chemical information and modeling, 57(7), 1563-1578 (2017-06-16)
Protein dynamics play a critical role in ligand binding, and different models have been proposed to explain the relationships between protein motion and molecular recognition. Here, we present a study of ligand-binding processes associated with large conformational changes of a
Thongkorn Ploypetchara et al.
Journal of food science, 85(10), 3372-3379 (2020-09-20)
The concentrations effects of sugars (trehalose and allose) and oils (canola and coconut oil) on the characteristics of rice starch suspension and mechanical properties of rice starch film were studied. The samples were prepared using 3% (w/w) rice starch, with
Dmitry A Rodionov et al.
BMC genomics, 11, 494-494 (2010-09-15)
Carbohydrates are a primary source of carbon and energy for many bacteria. Accurate projection of known carbohydrate catabolic pathways across diverse bacteria with complete genomes constitutes a substantial challenge due to frequent variations in components of these pathways. To address

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service