Skip to Content
Merck
All Photos(3)

Key Documents

274194

Sigma-Aldrich

Methyl 3,4,5-trihydroxybenzoate

98%

Synonym(s):

Methyl gallate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2CO2CH3
CAS Number:
Molecular Weight:
184.15
Beilstein:
2113180
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

mp

201-203 °C (lit.)

SMILES string

COC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C8H8O5/c1-13-8(12)4-2-5(9)7(11)6(10)3-4/h2-3,9-11H,1H3

InChI key

FBSFWRHWHYMIOG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rayza Morganna Farias Cavalcanti et al.
3 Biotech, 8(6), 270-270 (2018-06-06)
One of the tannase isoforms produced by the fungus Aspergillus fumigatus CAS-21 under submerged fermentation (SbmF) was purified 4.9-fold with a 10.2% recovery. The glycoprotein (39.1% carbohydrate content) showed an estimated molecular mass of 60 kDa. Optimum temperature and pH for
Judith Delius et al.
PloS one, 12(9), e0184487-e0184487 (2017-09-09)
Nuclear magnetic resonance (NMR) spectroscopy is well-established in assessing the binding affinity between low molecular weight ligands and proteins. However, conventional NMR-based binding assays are often limited to small proteins of high purity and may require elaborate isotopic labeling of
J P Chu et al.
Journal of dentistry, 35(5), 383-387 (2007-01-02)
To evaluate the effect of compounds of Galla chinensis on the remineralisation of initial enamel carious lesions in vitro. Sixty bovine enamel blocks with early lesions were prepared and randomly divided into six treatment groups. The lesions were subjected to
Boyun Sun et al.
Frontiers in pharmacology, 9, 1235-1235 (2018-11-16)
Ulcerative colitis (UC), with a long course and repeated attack, severely affects patient's life quality and increases economic burden all over the world. However, the concrete causes and mechanisms of UC are still unclear, but it is generally considered that
Deepika Singh et al.
Molecules (Basel, Switzerland), 24(4) (2019-02-20)
Leea indica (Vitaceae) is a Southeast Asian medicinal plant. In this study, an ethyl acetate fraction of L. indica leaves was studied for its phytoconstituents using high-performance liquid chromatography-electrospray ionization-mass spectrometry (HPLC-ESI-microTOF-Q-MS/MS) analysis. A total of 31 compounds of different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service