Skip to Content
Merck
All Photos(1)

Key Documents

266809

Sigma-Aldrich

Hafnium

turnings, crystal bar, 99.7% trace metals basis

Synonym(s):

Celtium, Hafnium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hf
CAS Number:
Molecular Weight:
178.49
MDL number:
UNSPSC Code:
12141718
PubChem Substance ID:
NACRES:
NA.23

Assay

99.7% trace metals basis

form

turnings, crystal bar

resistivity

29.6 μΩ-cm, 0°C

bp

4602 °C (lit.)

mp

2227 °C (lit.)

density

13.3 g/cm3 (lit.)

SMILES string

[Hf]

InChI

1S/Hf

InChI key

VBJZVLUMGGDVMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher C Underwood et al.
Inorganic chemistry, 52(1), 237-244 (2012-12-19)
This paper describes the hydrothermal chemistry of alkali hafnium fluorides, including the synthesis and structural characterization of five new alkali hafnium fluorides. Two ternary alkali hafnium fluorides are described: Li(2)HfF(6) in space group P31m with a = 4.9748(7) Å and
Yu Gong et al.
Dalton transactions (Cambridge, England : 2003), 41(38), 11706-11715 (2012-08-18)
The isolated group 4 metal oxydifluoride molecules OMF(2) (M = Ti, Zr, Hf) with terminal oxo groups are produced specifically on the spontaneous reactions of metal atoms with OF(2) through annealing in solid argon. The product structures and vibrational spectra
Byung-Hak Moon et al.
Journal of nanoscience and nanotechnology, 11(8), 7428-7432 (2011-11-23)
In this study, we investigated the electrochemical oxide nanotube formation on the Ti-35Ta-xHf alloys for dental materials. The Ti-35Ta-xHf alloys contained from 3 wt.% to 15 wt.% Hf were manufactured by arc melting furnace. The nanotube oxide layers were formed
Célia Lourenço et al.
The journal of physical chemistry. A, 116(51), 12399-12405 (2012-11-29)
Fourier transform ion cyclotron resonance mass spectrometry was used to characterize the gas-phase reactivity of Hf dipositive ions, Hf(2+)and HfO(2+), toward several oxidants: thermodynamically facile O-atom donor N(2)O, ineffective donor CO, and intermediate donors O(2), CO(2), NO, and CH(2)O. The
Laurence Maggiorella et al.
Future oncology (London, England), 8(9), 1167-1181 (2012-10-04)
There is considerable interest in approaches that could improve the therapeutic window of radiotherapy. In this study, hafnium oxide nanoparticles were designed that concentrate in tumor cells to achieve intracellular high-energy dose deposit. Conventional methods were used, implemented in different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service