Skip to Content
Merck
All Photos(2)

Key Documents

266000

Sigma-Aldrich

Lithium

greener alternative

ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis

Synonym(s):

Lithium foil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li
CAS Number:
Molecular Weight:
6.94
EC Number:
MDL number:
UNSPSC Code:
12141803
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

ribbon

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

9.446 μΩ-cm, 20°C

thickness × W

1.5 mm × 100 mm

bp

1342 °C (lit.)

mp

180 °C (lit.)

density

0.534 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li]

InChI

1S/Li

InChI key

WHXSMMKQMYFTQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium ribbon, thickness × W 1.5 mm × 100 mm is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium ribbon can be used as a counter and a reference electrode for lithium-ion batteries. It can also be used in the fabrication of textile-based batteries.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation on the solid electrolyte interface formed on pyrolyzed photoresist carbon anodes for C-MEMS lithium-ion batteries
Galobardes F, et al.
Diamond and Related Materials, 15(11-12), 1930-1934 (2006)
A layer-built rechargeable lithium ribbon-type battery for high energy density textile battery applications
Kim J, et al.
Journal of Material Chemistry A, 2(6), 1774-1780 (2014)
Enikö Zörgö et al.
PLoS genetics, 9(3), e1003388-e1003388 (2013-04-05)
The number of chromosome sets contained within the nucleus of eukaryotic organisms is a fundamental yet evolutionarily poorly characterized genetic variable of life. Here, we mapped the impact of ploidy on the mitotic fitness of baker's yeast and its never
Dongyun Chen et al.
Nanoscale, 5(17), 7890-7896 (2013-07-16)
Two-dimensional nanosheets can leverage on their open architecture to support facile insertion and removal of Li(+) as lithium-ion battery electrode materials. In this study, two two-dimensional nanosheets with complementary functions, namely nitrogen-doped graphene and few-layer WS2, were integrated via a
Jian Jiang et al.
Nanoscale, 5(17), 8105-8113 (2013-07-26)
Controlled integration of multiple semiconducting oxides into each single unit of ordered nanotube arrays is highly desired in scientific research for the realization of more attractive applications. We herein report a diffusion-controlled solid-solid route to evolve simplex Co(CO3)0.5(OH)0.11H2O@TiO2 core-shell nanowire

Articles

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service