Skip to Content
Merck
All Photos(2)

Key Documents

260789

Sigma-Aldrich

8-Aminoquinoline

98%

Synonym(s):

8-Quinolinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
Beilstein:
114474
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reaction type: C-H Activation
reagent type: catalyst

bp

174 °C/26 mmHg (lit.)

mp

60-65 °C (lit.)

SMILES string

Nc1cccc2cccnc12

InChI

1S/C9H8N2/c10-8-5-1-3-7-4-2-6-11-9(7)8/h1-6H,10H2

InChI key

WREVVZMUNPAPOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

8-Aminoquinoline fluoresce moderately to weakly in low dielectric media but not in strongly hydrogen-bonding or acidic aqueous media. The reaction of 8-aminoquinoline with chromium (III), manganese (II), iron (II) and (III), cobalt (II), nickel (II), copper (II), zinc (II), cadmium (II) and platinum (II) salts has been studied.

Application

8-Aminoquinoline has been used in:
  • preparation of base-stabilized terminal borylene complex of osmium
  • spectrophotometric determination of bivalent palladium

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Spectrophotometric Determination of Palladium with 8-Aminoquinoline.
Gustin VK and Sweet TR.
Analytical Chemistry, 35(1), 44-46 (1963)
Some transition metal complexes of 8-aminoquinoline.
Fanning JC and Taylor LT.
J. Inorg. Nucl. Chem., 27(10), 2217-2223 (1965)
Kyalo Stephen Kanyiva et al.
Organic letters, 16(7), 1968-1971 (2014-03-22)
A palladium-catalyzed regioselective activation of C(sp(2))-H and C(sp(3))-H bonds of benzamides and carboxamides, followed by coupling with disilanes to afford organosilanes in moderate to high yields, with the aid of 8-aminoquinoline as a directing group, is reported. Catalytic C(sp(2))-H germanylation
Hla Y Myint et al.
The American journal of tropical medicine and hygiene, 85(6), 1010-1014 (2011-12-07)
Because 8-aminoquinolines affect critical survival stages of Plasmodium parasites, treatment and control of malaria could be markedly improved by more widespread use of these drugs; however, hemolytic toxicity, which is widely prevalent in G6PD-deficient patients, severely constrains this use. Primaquine
Luis Carvalho et al.
Antimicrobial agents and chemotherapy, 55(9), 4204-4210 (2011-06-15)
The 8-aminoquinoline analogue sitamaquine (SQ) is an oral antileishmanial drug currently undergoing phase 2b clinical trials for the treatment of visceral leishmaniasis. In the present study, we investigated the mechanism of action of this drug in Leishmania donovani promastigotes. SQ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service