Skip to Content
Merck
All Photos(1)

Documents

260290

Sigma-Aldrich

1,2-Tetradecanediol

technical grade, 90%

Synonym(s):

1,2-Dihydroxytetradecane, 1,2-Tetradecylene glycol, Myristyl glycol, n-Tetradecane-1,2-diol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CH(OH)CH2OH
CAS Number:
Molecular Weight:
230.39
Beilstein:
1748243
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

Assay

90%

form

powder

mp

62-64 °C (lit.)

SMILES string

CCCCCCCCCCCCC(O)CO

InChI

1S/C14H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-14(16)13-15/h14-16H,2-13H2,1H3

InChI key

DWANEFRJKWXRSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Tong et al.
ACS nano, 11(7), 6808-6816 (2017-06-20)
The ability to generate heat under an alternating magnetic field (AMF) makes magnetic iron oxide nanoparticles (MIONs) an ideal heat source for biomedical applications including cancer thermoablative therapy, tissue preservation, and remote control of cell function. However, there is a
Diwei Ho et al.
Scientific reports, 7(1), 2866-2866 (2017-06-08)
Multimodal polymeric nanoparticles have many exciting diagnostic and therapeutic applications, yet their uptake and passage by the placenta, and applications in the treatment of pregnancy complications have not been thoroughly investigated. In this work, the maternal-fetal-placental biodistribution of anionic and
Jo J L Humphrey et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(36), 12694-12701 (2015-07-24)
Oleylamine (OA) based "hot injection" colloidal synthesis offers a versatile approach to the synthesis of highly monodisperse metallic and multi-metallic alloyed nanostructures in the absence of potentially toxic and unstable phosphine compounds. For application in heterogeneous catalysis and electrocatalysis, the

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service