Skip to Content
Merck
All Photos(1)

Key Documents

25580

Sigma-Aldrich

4-Chloro-2-nitrophenol

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)OH
CAS Number:
Molecular Weight:
173.55
Beilstein:
2048031
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

solid

impurities

~5% water

mp

85-87 °C (lit.)
85-89 °C

solubility

dioxane: soluble 0.5 g/5mL, clear, faintly yellow to yellow

SMILES string

Oc1ccc(Cl)cc1[N+]([O-])=O

InChI

1S/C6H4ClNO3/c7-4-1-2-6(9)5(3-4)8(10)11/h1-3,9H

InChI key

NWSIFTLPLKCTSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adsorption of 4-chloro-2-nitrophenol onto single-walled and multi-walled carbon nanotubes has been studied. Biotransformation of 4-chloro-2-nitrophenol by Bacillus sp. strain MW-1 has been studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gajendra Kumar Pradhan et al.
ACS applied materials & interfaces, 4(2), 707-713 (2011-12-29)
A series of nanostructure zinc-iron mixed oxide photocatalysts have been fabricated by solution-combustion method using urea as the fuel, and nitrate salts of both iron and zinc as the metal source. Different characterization tools, such as X-ray diffraction (XRD), X-ray
Degradation of trace aqueous 4-chloro-2-nitrophenol occurring in pharmaceutical industrial wastewater by ozone.
Gharbani P, et al.
International Journal of Environmental Science and Technology, 7(2), 377-384 (2010)
Pankaj Kumar Arora et al.
Biodegradation, 23(2), 325-331 (2011-09-06)
Decolourization, detoxification and biotransformation of 4-chloro-2-nitrophenol (4C2NP) by Bacillus sp. strain MW-1 were studied. This strain decolorized 4C2NP only in the presence of an additional carbon source. On the basis of thin layer chromatography (TLC), high performance liquid chromatography (HPLC)
Ali Mehrizad et al.
Iranian journal of environmental health science & engineering, 9(1), 5-5 (2013-02-02)
The adsorption characteristics of 4-chloro-2-nitrophenol (4C2NP) onto single-walled and multi-walled carbon nanotubes (SWCNTs and MWCNTs) from aqueous solution were investigated with respect to the changes in the contact time, pH of solution, carbon nanotubes dosage and initial 4C2NP concentration. Experimental
Babita Sur et al.
The Science of the total environment, 426, 296-303 (2012-04-24)
Anionic 2-nitro-4-chlorophenol (NCP) may occur in surface waters as a nitroderivative of 4-chlorophenol, which is a transformation intermediate of the herbicide dichlorprop. Here we show that NCP would undergo efficient photochemical transformation in environmental waters, mainly by direct photolysis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service