Skip to Content
Merck
All Photos(1)

Key Documents

252816

Sigma-Aldrich

Tetrahexylammonium bromide

99%

Synonym(s):

THABr

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5]4N(Br)
CAS Number:
Molecular Weight:
434.58
Beilstein:
3637302
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

99-100 °C (lit.)

SMILES string

[Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C24H52N.BrH/c1-5-9-13-17-21-25(22-18-14-10-6-2,23-19-15-11-7-3)24-20-16-12-8-4;/h5-24H2,1-4H3;1H/q+1;/p-1

InChI key

SYZCZDCAEVUSPM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetrahexylammonium bromide can be used:
  • As a hydrogen bond acceptor in the preparation of deep eutectic solvents which can be used for desulfurization and denitrogenation from n-heptane.
  • To produce aqueous biphasic system for the heavy metal ions preconcentration.
  • As a phase transfer salt to fabricate an optical chemical gas-sensor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multielement Determination of Trace Heavy Metals in Water by Microwave-Induced Plasma Atomic Emission Spectrometry after Extraction in Unconventional Single-Salt Aqueous Biphasic System
Smirnova SV, et al.
Analytical Chemistry, 90(10), 6323-6331 (2018)
Inkjet printed LED based pH chemical sensor for gas sensing
O?Toole M, et al.
Analytica Chimica Acta, 652(1-2), 308-314 (2009)
Extraction of benzothiazole and thiophene from their mixtures with n-heptane using tetrahexylammonium bromide-based deep eutectic solvents as extractive denitrogenation and desulfurization agents
Alli RD and Kroon MC
Fluid Phase Equilibria, 477, 1-11 (2018)
L Zhang et al.
The Journal of pharmacology and experimental therapeutics, 288(3), 1192-1198 (1999-02-23)
Polyspecific organic cation transporters in epithelia play an important role in the elimination of many endogenous bioactive amines and therapeutically important drugs. Recently, the first human organic cation transporter (hOCT1) was cloned from liver. The purpose of the current study
Y Uyama et al.
Pflugers Archiv : European journal of physiology, 426(5), 363-370 (1994-03-01)
Effects of tetraalkylammonium ions, having tetraalkyl chains of increasing length from ethyl to octyl, on inositol-trisphosphate (InsP3)-induced Ca2+ release and contractile mechanics were examined in guinea-pig skinned ileal smooth muscle longitudinal strips. Although tetrahexylammonium ions (THexA) appeared to be the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service