Skip to Content
Merck
All Photos(2)

Key Documents

252662

Sigma-Aldrich

Triethylaluminum solution

1.0 M in hexanes

Synonym(s):

Aluminumtriethanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(C2H5)3
CAS Number:
Molecular Weight:
114.16
Beilstein:
3587229
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.0 M in hexanes

density

0.692 g/mL at 25 °C

SMILES string

CC[Al](CC)CC

InChI

1S/3C2H5.Al/c3*1-2;/h3*1H2,2H3;

InChI key

VOITXYVAKOUIBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylaluminum solution can be used:
  • As a reagent in Alumino-Mannich reaction, where it reacts with iminium ion substrates for which the corresponding Petasis borono-Mannich reaction is unsuccessful.
  • In the synthesis of macrocyclic binuclear aluminum complexes with efficient activities toward ring-opening polymerization of ε-caprolactone.
  • To promote Claisen rearrangement of allyl vinyl ether derivatives along with the transfer of ethyl group to aldehydic carbon.
  • As a reagent for copper-catalyzed asymmetric conjugate addition of alkenyl- and alkylalanes to α,β-unsaturated lactams.
  • As a precursor to synthesize diethylaluminium cyanide, which is used as hydrocyanating reagent.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Self-heat. 2 - Skin Corr. 1A - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binuclear aluminum complexes with amine?imine type ligands derived from 1, 3-benzenedialdehyde: synthesis, structures and their catalytic properties in ring-opening polymerization.
Wu C, et al.
Journal of Coordination Chemistry, 69(6), 1066-1075 (2016)
Hydrocyanation. IV. New hydrocyanation methods using hydrogen cyanide and an alkylaluminum, and an alkylaluminum cyanide.
Nagata W, et al.
Journal of the American Chemical Society, 94(13), 4635-4643 (1972)
Aliphatic Claisen rearrangement promoted by organoaluminium reagents.
Takai K, et al.
Bulletin of the Chemical Society of Japan, 57(2), 446-451 (1984)
Copper-Catalyzed Asymmetric Conjugate Addition of Alkenyl-and Alkylalanes to ?, ?-Unsaturated Lactams.
Cottet P, et al.
Organic Letters, 15(4), 828-831 (2013)
An Alumino?Mannich Reaction of Organoaluminum Reagents, Silylated Amines, and Aldehydes.
Tarasewicz A, et al.
Chemistry?A European Journal , 24(23), 6071-6074 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service