Skip to Content
Merck
All Photos(1)

Key Documents

252336

Sigma-Aldrich

2,6-Dimethylphenyl isocyanate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NCO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.5356 (lit.)

bp

87-89 °C/12 mmHg (lit.)

density

1.057 g/mL at 25 °C (lit.)

functional group

isocyanate

storage temp.

2-8°C

SMILES string

Cc1cccc(C)c1N=C=O

InChI

1S/C9H9NO/c1-7-4-3-5-8(2)9(7)10-6-11/h3-5H,1-2H3

InChI key

YQLRKXVEALTVCZ-UHFFFAOYSA-N

Application

2,6-Dimethylphenyl isocyanate has been used in the preparation of:
  • derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography
  • tris( 2,6-dimethylphenylimido)methylrhenium (VII)
  • 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan Clayden et al.
Organic & biomolecular chemistry, 6(16), 2908-2913 (2008-08-09)
Except in the most hindered of cases, N,N'-diaryl N,N'-dimethyl ureas adopt a conformation with the two aryl rings disposed cis to one another. Variable temperature NMR studies reveal the rate at which the Ar-N bonds rotate as well as the
Selective substitution of an imido ligand in alkyl-and aryl-imido rhenium (VII) complexes; crystal structures of (2, 6-dimethylphenyl) ammonium trichlorobis (2, 6-dimethylphenyl-imido) methylrhenate (VII) and bis (2, 6-dimethylphenylimido) methylbis (neopentyl) rhenium (VII).
Cook MR, et al.
J. Chem. Soc., Dalton Trans., 797-804 (1991)
Derivatized cyclodextrins for normal-phase liquid chromatographic separation of enantiomers.
Armstrong DW, et al.
Analytical Chemistry, 62(15), 1610-1615 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service