Skip to Content
Merck
All Photos(1)

Key Documents

250287

Sigma-Aldrich

1,10-Diiododecane

95%

Synonym(s):

Decamethylene diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I(CH2)10I
CAS Number:
Molecular Weight:
394.07
Beilstein:
1738614
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

bp

197-200 °C/12 mmHg (lit.)

mp

33-35 °C (lit.)

SMILES string

ICCCCCCCCCCI

InChI

1S/C10H20I2/c11-9-7-5-3-1-2-4-6-8-10-12/h1-10H2

InChI key

CKJCTZAIDVFHCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Electrochemical reduction of 1,10-diiododecane at Hg cathode in dimethylformamide containing tetramethylammonium perchlorate has been investigated.

Application

1,10-Diiododecane has been used as:
  • an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes
  • crosslinking reagent for tetrabutylammonium polygalacturonic acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magali Darzac et al.
Chemical communications (Cambridge, England), (1)(1), 48-49 (2002-07-18)
After deprotection with a palladium catalyst, mono-allylated cryptophane-A (1, 2) and cryptophane-E (3) gave the new cryptophanols 4, 5 and 6, respectively, which are important key compounds for the preparation of monofunctionalized cryptophanes as well as for the design of
Electrochemical reduction of 1, 10-dihalodecanes at mercury cathodes in dimethylformamide.
Bart JC and Peters DG.
J. Electroanal. Chem. Interfac. Electrochem., 280(1), 129-144 (1990)
Gelation of chemically cross-linked polygalacturonic acid derivatives.
Matricardi P, et al.
Carbohydrate Polymers, 27(3), 215-220 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service