Skip to Content
Merck
All Photos(1)

Documents

246425

Sigma-Aldrich

trans,trans-Dibenzylideneacetone

98%

Synonym(s):

1,5-Diphenyl-1,4-pentadien-3-one, trans,trans-1,5-Diphenyl-1,4-pentadien-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOCH=CHC6H5
CAS Number:
Molecular Weight:
234.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

104-107 °C (lit.)

solubility

acetone: soluble(lit.)
alcohol: slightly soluble(lit.)
chloroform: soluble(lit.)
diethyl ether: slightly soluble(lit.)
water: insoluble(lit.)

SMILES string

O=C(\C=C\c1ccccc1)/C=C/c2ccccc2

InChI

1S/C17H14O/c18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16/h1-14H/b13-11+,14-12+

InChI key

WMKGGPCROCCUDY-PHEQNACWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans,trans-Dibenzylideneacetone was used as an additive in the copper-catalyzed N-arylation of imidazoles.
Reactant involved in:
  • Nazarov-like cyclization
  • Transfer hydrogenation
  • Lewis acid mediated condensation
  • Hetero-Diels-Alder reactions
  • Asymmetric 1,4-addition reactions
  • Michael addition reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rahul Balasaheb Aher et al.
Bioorganic & medicinal chemistry letters, 21(10), 3034-3036 (2011-04-16)
A series of dibenzylideneacetones (A1-A12) and some of their pyrazolines (B1-B4) were synthesized and evaluated in vitro for blood stage antiplasmodial properties in Plasmodium falciparum culture using SYBR-green-I fluorescence assay. The compound (1E, 4E)-1,5-bis(3,4-dimethoxyphenyl)penta-1,4-dien-3-one (A9) was found to be the
Kiyomori A, et al.
Tetrahedron Letters, 40(14), 2657-2660 null
Regina Appiah-Opong et al.
European journal of medicinal chemistry, 43(8), 1621-1631 (2008-02-06)
Inhibition of cytochrome P450 (CYP) is a major cause of drug-drug interactions. In this work, inhibitory potentials of 33 curcumin analogues, i.e. 2,6-dibenzylidenecyclohexanone (A series), 2,5-dibenzylidenecyclopentanone (B series) and 1,4-pentadiene-3-one (C series) substituted analogues of curcumin towards recombinant human CYP1A2
D Batovska et al.
European journal of medicinal chemistry, 42(1), 87-92 (2006-09-30)
A large series of chalcones were synthesized and studied for activity against Candida albicans. The SAR analysis showed that the antifungal activity was highly dependent on the substitution pattern of the aryl rings and correlated to a large extent with
Mengchao Cui et al.
Journal of medicinal chemistry, 54(7), 2225-2240 (2011-03-23)
A new and extensive set of dibenzylideneacetone derivatives was synthesized and screened for affinity toward Aβ(1-42) aggregates. Structure-activity relationships revealed the binding of dibenzylideneacetones to be affected by various substituents. The introduction of a substituent group in the ortho position

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service