Skip to Content
Merck
All Photos(1)

Documents

242837

Sigma-Aldrich

2-Bromohexanoic acid

97%

Synonym(s):

2-Bromocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CHBrCOOH
CAS Number:
Molecular Weight:
195.05
Beilstein:
1721762
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.472 (lit.)

bp

136-138 °C/18 mmHg (lit.)

solubility

alcohol: soluble(lit.)
diethyl ether: soluble(lit.)

density

1.37 g/mL at 25 °C (lit.)

SMILES string

CCCCC(Br)C(O)=O

InChI

1S/C6H11BrO2/c1-2-3-4-5(7)6(8)9/h5H,2-4H2,1H3,(H,8,9)

InChI key

HZTPKMIMXLTOSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The reaction of 2-bromohexanoic acid (2-bromocaproic acid) with 6-(5,6-dipentyl-1,2,4-triazin-3-yl)-2,2′-bipyridine and Cm(III) was studied.

Application

2-Bromohexanoic acid was used as an internal standard in the determination of trans,trans-muconic acid (t,t-MA) in urine by GC-MS method.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ruppert et al.
Journal of chromatography. B, Biomedical applications, 666(1), 71-76 (1995-04-07)
A sensitive and specific method for the determination of trans,trans-muconic acid (t,t-MA) in urine is described. After clean-up on an anion-exchange cartridge, t,t-MA was derivatized with BF3-methanol to the dimethyl ester and analyzed by gas chromatography-mass spectrometry (GC-MS), with 2-bromohexanoic
Antje Bremer et al.
Dalton transactions (Cambridge, England : 2003), 41(25), 7582-7589 (2012-05-19)
The complexation of Cm(III) with 6-(5,6-dipentyl-1,2,4-triazin-3-yl)-2,2'-bipyridine (C(5)-hemi-BTP) in a water-2-propanol solution is investigated by TRLFS. Upon increasing the concentration of C(5)-hemi-BTP in the Cm(III)-solution, three different species, the 1:1-, 1:2- and 1:3-complex, with emission bands at 599.9 nm, 607.3 nm
Damien E Dobson et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 19(1), 56-65 (2019-12-12)
A series of Cy5.5 dye analogs and targeted probes with net charges varied from -3 to 0 were synthesized by an optimized method, followed by comparing their spectral and photostability properties in saturated solutions of air, oxygen, and argon. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service