Skip to Content
Merck
All Photos(1)

Key Documents

241881

Sigma-Aldrich

Tungsten(IV) carbide

powder, 2 μm, ≥99%

Synonym(s):

Tungsten carbide, Tungsten monocarbide, Wolfram carbide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WC
CAS Number:
Molecular Weight:
195.85
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99%

form

powder

particle size

2 μm

SMILES string

[C-]#[W+]

InChI

1S/C.W/q-1;+1

InChI key

UONOETXJSWQNOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya Yan et al.
Small (Weinheim an der Bergstrasse, Germany), 8(21), 3350-3356 (2012-07-26)
A unique nanorod-structured tungsten carbide material with high specific surface area of 198 m(2) g(-1) is successfully synthesized for the first time by pseudomorphic transformation of chemically synthesized WO(3) nanorods through a high-temperature method. An electrocatalyst composed of Pt nanoparticles
Farzaneh Ahrari et al.
Progress in orthodontics, 13(1), 17-22 (2012-05-16)
There are controversial reports regarding the effect of repeated bonding on shear bond strength (SBS) of orthodontic attachments. The aim of this study was to evaluate the SBS of brackets following early and delayed rebonding, and after employing different methods
Dehong Shan et al.
Toxicology letters, 209(2), 129-135 (2011-12-24)
The effects of tungsten carbide nanoparticles (nano-WC) on the properties of voltage-dependent potassium currents and evoked action potentials were studied in the hippocampal CA1 pyramidal neurons of rats at the ages of postnatal days 10-14 using the whole-cell patch-clamp technique.
Na Ji et al.
ChemSusChem, 5(5), 939-944 (2012-04-03)
A series of Ni-promoted W(2) C catalysts was prepared by means of a post-impregnation method and evaluated for the catalytic conversion of cellulose into ethylene glycol (EG). Quite different from our previously reported Ni-W(2) C/AC catalysts, which were prepared by
Alan Islas-Cital et al.
The Journal of the Acoustical Society of America, 130(4), 1880-1887 (2011-10-07)
The phase angle component of the complex frequency response of a sonar system operating near transducer resonance is usually distorted. Interpretation and classification of the received sonar signal benefits from the preservation of waveform fidelity over the full bandwidth. A

Articles

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service