Skip to Content
Merck
All Photos(1)

Documents

234494

Sigma-Aldrich

1-Eicosanol

98%

Synonym(s):

Arachinyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19OH
CAS Number:
Molecular Weight:
298.55
Beilstein:
1705104
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.4 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

98%

mp

62-65 °C (lit.)

solubility

water: slightly soluble 0.001 g/L at 23 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCO

InChI

1S/C20H42O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h21H,2-20H2,1H3

InChI key

BTFJIXJJCSYFAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The polymorphism of 1-eicosanol has been studied by X-ray powder diffraction, differential scanning calorimetry (DSC), Raman scattering and IR spectroscopy.

Application

1-Eicosanol has been used:
  • in gradient HPLC-charged aerosol detection method for the detection of different lipids
  • as internal standard during determination of policosanol′s high molecular weight alcohols (policosanol′s fatty alcohols) in fish oil by GC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

381.2 °F - open cup

Flash Point(C)

194 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Géraldine Giacinti et al.
Journal of chromatography. A, 1483, 8-19 (2017-01-11)
The sample matrix can enhance the gas chromatography signal of pesticide residues relative to that obtained with the same concentration of pesticide in solvent. This paper is related to negative matrix effects observed in coupled gas chromatography-mass spectrometry ion trap
Polymorphism of n-alkanols: 1-Heptadecanol, 1-octadecanol, 1-nonadecanol, and 1-eicosanol.
Ventola L, et al.
Chemistry of Materials, 14(2), 508-517 (2002)
Marc Plante et al.
Methods in molecular biology (Clifton, N.J.), 579, 469-482 (2009-09-19)
A gradient HPLC-charged aerosol detection method was applied to the measurement of different lipids including: free fatty acids, fatty alcohols, glycerides, steroids, phospholipids, and fat-soluble vitamins. An algal oil sample is used as an example. Charged aerosol detection offers a
Ernesto J Méndez Antolín et al.
Journal of AOAC International, 91(5), 1013-1019 (2008-11-05)
A new validated gas chromatographic (GC) method using acetyl chloride as a derivatizing agent for determining policosanol's high molecular weight alcohols (policosanol's fatty alcohols; PFAs) in fish oil (FO) is described. Before derivatizing the alcohols, FO was subjected to a
Agnieszka Serafin et al.
The Journal of membrane biology, 248(4), 767-773 (2015-03-25)
Monolayers of phosphatidylcholine (PC), tetradecanol (TD), hexadecanol (HD), octadecanol (OD) and eicosanol (E) and their binary mixtures were investigated at the air/water interface. The surface tension values of pure and mixed monolayers were used to calculate π-A isotherms. The surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service