Skip to Content
Merck
All Photos(2)

Key Documents

232254

Sigma-Aldrich

1-Cyclohexyl-2-pyrrolidone

High-boiling aprotic solvent., 99%

Synonym(s):

1-Cyclohexyl-2-pyrrolidinone, CHP, N-Cyclohexyl-2-pyrrolidinone, N-Cyclohexyl-2-pyrrolidone, N-Cyclohexylpyrrolidinone, N-Cyclohexylpyrrolidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO
CAS Number:
Molecular Weight:
167.25
Beilstein:
121832
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.05 mmHg ( 25 °C)

Assay

99%

refractive index

n20/D 1.499 (lit.)

bp

154 °C/7 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCN1C2CCCCC2

InChI

1S/C10H17NO/c12-10-7-4-8-11(10)9-5-2-1-3-6-9/h9H,1-8H2

InChI key

PZYDAVFRVJXFHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

285.8 °F - closed cup

Flash Point(C)

141 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction of aromatics from petroleum naphtha reformate by a 1-cyclohexyl-2-pyrrolidone/ethylene carbonate mixed solvent.
Radwan GM, et al.
Industrial & Engineering Chemistry Research, 36(2), 414-418 (1997)
Karina Milagros Cui et al.
ACS applied materials & interfaces, 3(7), 2300-2308 (2011-06-10)
The facile preparation of poly (N-vinyl carbazole) (PVK) and multiwalled carbon nanotubes (MWNTs) solution and conjugated polymer network (CPN) nanocomposite film is described. The stable solutions of PVK/MWNT were prepared in mixed solvents by simple sonication method, which enabled successful
P J Becci et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 4(4), 587-593 (1984-08-01)
Teratogenesis studies were performed in rats and rabbits given N-cyclohexyl-2-pyrrolidone, an aprotic solvent used in chemical processing. Dosages of 0, 15, 50, 150, or 500 mg N-cyclohexyl-2-pyrrolidone/kg body wt/day were administered by gavage to groups of 25 pregnant Sprague-Dawley rats
E Q Lawson et al.
The Journal of biological chemistry, 259(5), 2910-2912 (1984-03-10)
The compound N-cyclohexyl-2-pyrrolidone contains a substantial apolar region as well as a peptide bond-like moiety. This solvent, therefore, provides a useful model for protein interiors. Under certain conditions of temperature and salt concentration, cyclohexylpyrrolidone forms a two-phase system with water.
Lothar Schwarz et al.
Journal of forensic sciences, 52(3), 649-655 (2007-04-26)
A new method for enhancement of ninhydrin or 1,8-diazafluoren-9-one (DFO)-treated latent fingerprints on thermal paper will be described. Most thermosensitive surfaces of thermal paper become dark when treated with DFO or ninhydrin petroleum ether (NPB) solution. This effect minimizes contrast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service