226076
Benzaldehyde dimethyl acetal
99%
Synonym(s):
α,α-Dimethoxytoluene
About This Item
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.493 (lit.)
bp
87-89 °C/18 mmHg (lit.)
density
1.014 g/mL at 25 °C (lit.)
SMILES string
COC(OC)c1ccccc1
InChI
1S/C9H12O2/c1-10-9(11-2)8-6-4-3-5-7-8/h3-7,9H,1-2H3
InChI key
HEVMDQBCAHEHDY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Benzaldehyde dimethyl acetal is derived from the condensation reaction between benzaldehyde and methanol. It is used as a protecting group.
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
156.2 °F - closed cup
Flash Point(C)
69 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service