224189
4-(Fluorosulfonyl)benzoic acid
95%
Synonym(s):
4-Carboxybenzenesulfonyl fluoride, p-(Fluorosulfonyl)benzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
FSO2C6H4CO2H
CAS Number:
Molecular Weight:
204.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
solid
reaction suitability
reaction type: click chemistry
mp
272-273 °C (lit.)
functional group
carboxylic acid
SMILES string
OC(=O)c1ccc(cc1)S(F)(=O)=O
InChI
1S/C7H5FO4S/c8-13(11,12)6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)
InChI key
DJUJJHDCOUPERR-UHFFFAOYSA-N
Related Categories
General description
4-(Fluorosulfonyl)benzoic acid is a a xenobiotic substrate analogue which on incubation with 4-4 isozyme of rat liver glutathione S-transferase results in a time-dependent inactivation of the enzyme.
Application
4-(Fluorosulfonyl)benzoic acid was used as an affinity label of dimeric pig lung pi class glutathione S-transferase.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lisa L Moore et al.
Journal of proteome research, 3(6), 1184-1190 (2004-12-15)
Proteins that bind ATP and GTP are important cellular components. We developed an immunological approach to selectively tag nucleotide-binding proteins based on the use of 5'-[4-(fluorosulfonyl)benzoyl]adenosine and 5'-[4-(fluorosulfonyl)benzoyl]guanosine affinity tags and an antibody against 4-(sulfonyl)benzoate. Detection follows affinity labeling, gel
N E Pettigrew et al.
Archives of biochemistry and biophysics, 364(1), 107-114 (1999-03-24)
The compound 4-(fluorosulfonyl)benzoic acid (4-FSB) functions as an affinity label of the dimeric pig lung pi class glutathione S-transferase yielding a completely inactive enzyme. Protection against inactivation is provided by glutathione-based ligands, suggesting that the reaction target is near or
J J Barycki et al.
Biochemistry, 32(48), 13002-13011 (1993-12-07)
Incubation of 4-(fluorosulfonyl)benzoic acid (4-FSB), a xenobiotic substrate analogue, with the 4-4 isozyme of rat liver glutathione S-transferase at pH 7.5 and 25 degrees C results in a time-dependent inactivation of the enzyme. The rate of inactivation exhibits a nonlinear
J Wang et al.
Protein science : a publication of the Protein Society, 5(6), 1032-1042 (1996-06-01)
Reaction of rat liver glutathione S-transferase, isozyme 1-1, with 4-(fluorosulfonyl)benzoic acid (4-FSB), a xenobiotic substrate analogue, results in a time-dependent inactivation of the enzyme to a final value of 35% of its original activity when assayed at pH 6.5 with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service