Skip to Content
Merck
All Photos(1)

Key Documents

223220

Sigma-Aldrich

Thiocarbohydrazide

98%

Synonym(s):

Thiocarbonyldihydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2NH)2CS
CAS Number:
Molecular Weight:
106.15
Beilstein:
506657
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39093513
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

171-174 °C (dec.) (lit.)

functional group

amine
hydrazine
thiourea

SMILES string

NNC(=S)NN

InChI

1S/CH6N4S/c2-4-1(6)5-3/h2-3H2,(H2,4,5,6)

InChI key

LJTFFORYSFGNCT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Review of transition metal complexes with thiocarbohydrazides: This comprehensive review discusses the coordination chemistry of thiocarbohydrazides with various metals, highlighting their relevance in the synthesis of complex metal compounds used in catalysis and pharmaceutical research (Aly et al., 2023).

  • Pd-doped nanocomposites for organometallic catalysis: Thiocarbohydrazide was employed in the fabrication of Pd-doped SBA-15 nanocomposites, applied as catalysts in the synthesis of organometallic compounds, showcasing its utility as a catalyst support material (Kalhor and Dadras, 2021).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamaleddin Haj Mohammad Ebrahim Tehrani et al.
Iranian journal of pharmaceutical research : IJPR, 12(2), 331-346 (2013-11-20)
In this work, we reported the synthesis and evaluation of antimycobacterial and antifungal activity of a series of thiocarbohydrazone derivatives which are thiacetazone congeners. The target compounds were synthesized in superior yields by reacting thiocarbohydrazide with different aromatic aldehydes and
D Levanon et al.
The Histochemical journal, 31(1), 71-73 (1999-07-16)
Samples from seven sectors of the rabbit knee articular cartilage were shaved and prepared for the scanning electron microscope using either tannic acid, thiocarbohydrazide or nothing (control). Surface morphology was found to be more typical to a given sector and
D P Singh et al.
Journal of enzyme inhibition and medicinal chemistry, 22(2), 177-182 (2007-05-24)
A novel series of complexes of the type [M(TML)X2]; where TML is Tetradentate Macrocyclic Ligand; M = Co(II), Ni(II), Cu(II), Zn(II)or Cd(II); X = Cl-, CH3COO- or NO3- have been synthesized by template condensation of glyoxal and thiocarbohydrazide in the
Debabrata Maity et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(40), 11152-11161 (2011-09-02)
We present novel Schiff base ligands julolidine-carbonohydrazone 1 and julolidine-thiocarbonohydrazone 2 for selective detection of Cu(2+) in aqueous medium. The planar julolidine-based ligands can sense Cu(2+) colorimetrically with characteristic absorbance in the near-infrared (NIR, 700-1000 nm) region. Employing molecular probes
M Goldberg et al.
European journal of oral sciences, 104(3), 269-277 (1996-06-01)
The appearance of the junction between predentin and dentin of rat incisors was investigated after chemical fixation (aldehyde), physical fixation (high pressure freezing and freeze substitution) and by histochemistry. Physical fixation revealed a 1-2 micron wide intermediary zone, in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service