Skip to Content
Merck
All Photos(1)

Key Documents

220116

Sigma-Aldrich

2-Hydroxybutyric acid sodium salt

97%

Synonym(s):

2-Hydroxybutanoic acid sodium salt, DL-α-Hydroxybutyric acid sodium salt, Sodium α-hydroxybutyrate, Sodium (DL)-α-hydroxybutyrate, Sodium (±)-2-hydroxybutyrate, Sodium 2-hydroxybutanoate, Sodium 2-hydroxybutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(OH)CO2Na
CAS Number:
Molecular Weight:
126.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

133-135 °C (lit.)

functional group

hydroxyl

SMILES string

[Na+].CCC(O)C([O-])=O

InChI

1S/C4H8O3.Na/c1-2-3(5)4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1

InChI key

MOSCXNXKSOHVSQ-UHFFFAOYSA-M

Application

2-Hydroxybutyric acid sodium salt was used in gas chromatographic (GC) method for the indirect enantioresolution of 2- and 3-hydroxy fatty acids[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramona Jenske et al.
Journal of chromatography. A, 1146(2), 225-231 (2007-02-13)
A gas chromatographic (GC) method is described for the indirect enantioresolution of 2- and 3-hydroxy fatty acids (OH-FAs). It combines the derivatization of each alkylated enantiomer and the subsequent transfer with (R)-(-)-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride [(R)-(-)-MTPA-Cl, Mosher's reagent] into a diastereomeric (S)-MTPA
Rawi Ramautar et al.
Analytical and bioanalytical chemistry, 387(1), 293-301 (2006-11-11)
Organic acids in cerebrospinal fluid (CSF) are potential diagnostic markers for neurological diseases and metabolic disorders. A capillary electrophoretic (CE) method for the direct analysis, i.e., without any sample preparation, of six organic acids in CSF was developed. A capillary
Ele Ferrannini et al.
Diabetes, 62(5), 1730-1737 (2012-11-20)
Metabolomic screening of fasting plasma from nondiabetic subjects identified α-hydroxybutyrate (α-HB) and linoleoyl-glycerophosphocholine (L-GPC) as joint markers of insulin resistance (IR) and glucose intolerance. To test the predictivity of α-HB and L-GPC for incident dysglycemia, α-HB and L-GPC measurements were
Shihhui Huang et al.
Hippocampus, 25(8), 963-975 (2015-01-24)
Work on laboratory and wild rodents suggests that domestication may impact on the extent of adult hippocampal neurogenesis and its responsiveness to regulatory factors. There is, however, no model of laboratory rodents and their nondomesticated conspecifics that would allow a
Konrad Hoetzenecker et al.
European heart journal, 36(11), 676-685 (2013-01-17)
Supernatants of serum-free cultured mononuclear cells (MNC) contain a mix of immunomodulating factors (secretome), which have been shown to attenuate detrimental inflammatory responses following myocardial ischaemia. Inflammatory dilated cardiomyopathy (iDCM) is a common cause of heart failure in young patients.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service