Skip to Content
Merck
All Photos(1)

Key Documents

219355

Sigma-Aldrich

D-Gulonic acid γ-lactone

97%

Synonym(s):

D-(−)-Gulono-1,4-lactone, D-Gulono γ-lactone, D-Gulono-1,4-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Beilstein:
83012
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −55°, c = 4 in H2O

mp

182-188 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H]1OC(=O)[C@H](O)[C@@H]1O

InChI

1S/C6H10O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-5,7-10H,1H2/t2-,3+,4-,5+/m1/s1

InChI key

SXZYCXMUPBBULW-LECHCGJUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Braun et al.
FEBS letters, 388(2-3), 173-176 (1996-06-17)
The relationship between glutathione deficiency, glycogen metabolism and ascorbate synthesis was investigated in isolated murine hepatocytes. Glutathione deficiency caused by various agents increased ascorbate synthesis with a stimulation of glycogen breakdown. Increased ascorbate synthesis from UDP-glucose or gulonolactone could not
A Krasnov et al.
Genetic analysis : biomolecular engineering, 15(3-5), 115-119 (1999-12-22)
The reviewed studies addressed the possibility of using gene transfer for correction of L-ascorbic acid biosynthesis and carbohydrate utilization in rainbow trout. Analyses of enzymatic activities in the L-AAB pathway indicated that reasons for the lack of L-AA production can
F Puskás et al.
FEBS letters, 430(3), 293-296 (1998-08-04)
The orientation of gulonolactone oxidase activity was investigated in rat liver microsomes. Ascorbate formation upon gulonolactone addition resulted in higher intravesicular than extravesicular ascorbate concentrations in native microsomal vesicles. The intraluminal ascorbate accumulation could be prevented or the accumulated ascorbate
Purification and characterization of L-gulonolactone oxidase from chicken kidney microsomes.
K Kiuchi et al.
Biochemistry, 21(20), 5076-5082 (1982-09-28)
Yeryung Kim et al.
Journal of Korean medical science, 26(3), 317-324 (2011-03-12)
Hyperoxic ventilation induces detrimental effects on the respiratory system, and ambient oxygen may be harmful unless compensated by physiological anti-oxidants, such as vitamin C. Here we investigate the changes in electrolyte transport of airway epithelium in mice exposed to normobaric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service