Skip to Content
Merck
All Photos(2)

Key Documents

217891

Sigma-Aldrich

Nickel(II) bromide

98%

Synonym(s):

Nickel(2+) dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiBr2
CAS Number:
Molecular Weight:
218.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
solubility:
H2O: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

grade

for analytical purposes

Assay

98%

form

solid

reaction suitability

reagent type: catalyst
core: nickel

mp

963 °C (lit.)

solubility

H2O: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

density

5.098 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Br[Ni]Br

InChI

1S/2BrH.Ni/h2*1H;/q;;+2/p-2

InChI key

IPLJNQFXJUCRNH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam F Nugraha et al.
Polymers, 12(7) (2020-07-24)
Several methods to synthesize poly(phenylene) block copolymers through the nickel coupling reaction were attempted to reduce the use of expensive nickel catalysts in polymerization. The model reaction for poly(phenylene) having different types of dichlorobenzene derivative monomers illustrated the potential use

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service