Skip to Content
Merck
All Photos(1)

Documents

21604

Sigma-Aldrich

2-Bromohexadecanoic acid

≥99.0% (GC)

Synonym(s):

2-Bromopalmitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH(Br)CO2H
CAS Number:
Molecular Weight:
335.32
Beilstein:
1726517
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

form

solid

mp

52-54 °C

solubility

methanol: soluble 1 g/10 mL, clear, colorless
chloroform: soluble
ethanol: soluble
water: insoluble

SMILES string

CCCCCCCCCCCCCCC(Br)C(O)=O

InChI

1S/C16H31BrO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(17)16(18)19/h15H,2-14H2,1H3,(H,18,19)

InChI key

DPRAYRYQQAXQPE-UHFFFAOYSA-N

Gene Information

human ... PPARD(5467)

Looking for similar products? Visit Product Comparison Guide

General description

2-Bromohexadecanoic acid, also known as 2-Bromopalmitic acid, is a long chain 2-haloalkanoic halogenated fatty acid that is commonly used as a building block in various organic reaction such as 2′-(2-bromohexadecanoyl)-docetaxel synthesis.

Application

<ul>
<li><strong>Chemo-immunotherapy Applications:</strong> 2-Bromohexadecanoic acid has been utilized to create bioactive fatty acid analog-derived hybrid nanoparticles for chemo-immunotherapy against carcinoma, demonstrating potential in cancer treatment without the need for antibodies (Tan et al., 2023).</li>
<li><strong>Drug Target Identification for Coronavirus:</strong> 2-bromopalmitate (2-BP) can be used as an inhibitor and is used to suppressed SADS-CoV treatment (Luo et al., 2021).</li>
<li><strong>Cancer Therapy Enhancement:</strong> This compound has shown efficacy in sensitizing osteosarcoma cells to adriamycin-induced apoptosis through modulation of CHOP, indicating its therapeutic potential in enhancing cancer treatment (Xu et al., 2019).</li>
<li><strong>Membrane Protein Localization in Sertoli Cells: </strong>It has been involved in studies related to the localization of the androgen receptor to plasma membranes by binding to caveolin-1 in mouse Sertoli cells, contributing to the understanding of cellular signaling pathways. It is Used to inhibit palmitoylation of androgen receptor (AR) and used to evaluate the effect of the inhibitor on androgen receptor (AR) translocation (Deng et al., 2017).</li>
<li><strong>Palmitoylation Inhibition:</strong> 2-Bromohexadecanoic acid has been profiled as an irreversible inhibitor of palmitoylation, providing insights into the regulation of protein modifications and their implications in various diseases (Davda et al., 2013).</li>
</ul>

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibacterial and biofilm prevention metabolites from Acanthophora spicifera
Fitri B, et al.
open chemistry, 21, 20230163-20230163 (2023)
Molecular Dynamics Simulation Studies of Self-Assembly of Racemic (R)/(S)-2-Bromohexadecanoic Acid on a Graphite Surface: Enantio-pure or Enantio-mixed Domains?.
Ilan B, et al.
The Journal of Physical Chemistry C, 111(49), 18243-18250 (2007)
Oil-Filled Lipid Nanoparticles Containing 2?-(2-Bromohexadecanoyl)-Docetaxel for the Treatment of Breast Cancer
Lan F, et al.
Advanced Healthcare Materials, 2, 1451-1457 (2013)
I Nagase et al.
FEBS letters, 461(3), 319-322 (1999-11-24)
Uncoupling protein 3 (UCP3), expressed abundantly in the skeletal muscle, is one of the carrier proteins dissipating the transmitochondrial electrochemical gradient as heat, and thereby has been implicated in the regulation of energy metabolism. We have investigated UCP3 mRNA expression
Jarrod R Fortwendel et al.
Eukaryotic cell, 11(8), 966-977 (2012-05-09)
Ras is a highly conserved GTPase protein that is essential for proper polarized morphogenesis of filamentous fungi. Localization of Ras proteins to the plasma membrane and endomembranes through posttranslational addition of farnesyl and palmitoyl residues is an important mechanism through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service