All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H3IN2
CAS Number:
Molecular Weight:
193.97
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
solid
mp
108-110 °C (lit.)
functional group
iodo
SMILES string
Ic1cn[nH]c1
InChI
1S/C3H3IN2/c4-3-1-5-6-2-3/h1-2H,(H,5,6)
InChI key
LLNQWPTUJJYTTE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-Iodopyrazole is a valuable intermediate for the synthesis of biologically active compounds. It undergoes iodination in the presence of iodine and ammonium hydroxide to yield 3,4-di-iodo- and 3,4,5-tri-iodo-pyrazole.
Application
4-Iodopyrazole was used in an indium-mediated synthesis of heterobiaryls.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Enrique Font-Sanchis et al.
The Journal of organic chemistry, 72(9), 3589-3591 (2007-04-05)
The palladium-mediated coupling reaction between triorganoindium reagents and organic electrophiles is extended to the synthesis of heteroaromatic compounds. Both electron-rich and electron-poor heterocycles can act as the organic electrophile or as the organoindium derivative.
Green iodination of pyrazoles with iodine/hydrogen peroxide in water.
Kim MM, et al.
Tetrahedron Letters, 49(25), 4026-4028 (2008)
Some iodinated pyrazole derivatives.
D Giles et al.
Journal of the Chemical Society. Perkin transactions 1, 13, 1179-1184 (1966-01-01)
A Kojo et al.
Archives of toxicology, 72(6), 336-341 (1998-07-10)
Mouse liver CYP2A5 is induced by several structurally unrelated compounds. In intact mouse liver, pyrazole (PYR) and 4-hydroxypyrazole (4-OH) induce selectively the expression of CYP2A5 while expression of other CYPs is decreased. In this study we exposed mouse primary hepatocytes
Hsuan-Liang Liu et al.
Journal of biomedical science, 10(3), 302-312 (2003-04-25)
Molecular docking simulations were performed in this study to investigate the importance of both structural and catalytic zinc ions in the human alcohol dehydrogenase beta(2)beta(2) on substrate binding. The structural zinc ion is not only important in maintaining the structural
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service