Skip to Content
Merck
All Photos(1)

Key Documents

209708

Sigma-Aldrich

Lead

powder, −325 mesh, ≥99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
EC Number:
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99% trace metals basis

form

powder

resistivity

20.65 μΩ-cm

particle size

−325 mesh

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenyi Yuan et al.
Journal of the Air & Waste Management Association (1995), 63(4), 418-423 (2013-05-22)
A novelprocess ofmechanochemical sulfidization for the treatment of cathode ray tube (CRT)funnel glass has been investigated by co-grinding with the element sulfur The ground samples were characterized by means of a set of analytical methods, including scanning electron microscopy (SEM)
James D Blando et al.
Journal of environmental health, 75(9), 20-27 (2013-06-06)
The U.S. Environmental Protection Agency recently implemented the Renovation, Repair and Painting (RRP) rule that applies to pre-1978 residences because of the potential presence of lead-based paint. Enforcement of this rule may be difficult and therefore it is crucial to
Carla Campbell et al.
American journal of public health, 103(7), 1271-1277 (2013-05-18)
We determined whether Philadelphia Lead Court is effective in enforcing lead hazard remediation in the homes of children with elevated blood lead levels. We created a deidentified data set for properties with an initial failed home inspection (IFHI) for lead
Kai-Ying Chiang et al.
Journal of environmental sciences (China), 25(3), 626-636 (2013-08-08)
This study investigates biogeochemical reductive release of arsenate from beudantite into solution in a crater area in northern Taiwan, using a combination of X-ray absorption near-edge structure (XANES) and atomic absorption spectrometry. Total arsenic (As) concentrations in the soil were
Karsten Witt et al.
Brain : a journal of neurology, 136(Pt 7), 2109-2119 (2013-06-27)
Deep brain stimulation of the subthalamic nucleus improves motor functions in patients suffering from advanced Parkinson's disease but in some patients, it is also associated with a mild decline in cognitive functioning about one standard deviation from the preoperative state.

Articles

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service