Skip to Content
Merck
All Photos(3)

Documents

208515

Sigma-Aldrich

Niobium(V) oxide

−325 mesh, 99.9% trace metals basis

Synonym(s):

Niobium pentoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb2O5
CAS Number:
Molecular Weight:
265.81
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

particle size

−325 mesh

density

4.47 g/mL at 25 °C (lit.)

SMILES string

O=[Nb](=O)O[Nb](=O)=O

InChI

1S/2Nb.5O

InChI key

ZKATWMILCYLAPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Niobium(V) oxide, also known as niobium pentoxide, is a white crystalline powder. It is a refractory metal oxide with a high melting point of over 2500 °C and high chemical stability, including to oxygen and moisture. Niobium(V) oxide is mainly used as a raw material in the production of niobium and niobium alloys, as a catalyst in chemical reactions, and as a refractory material in high-temperature applications. It is also used as a dielectric material in capacitors and as a pigment in ceramics and glasses.

Application

Applied in the solid state formation of an unusual cation deficient perovskite, Ba7Nb4MoO20.
Promising results were obtained using niobium(V) oxide as an alternate electrode to lithium metal in advanced fuel cells.

Features and Benefits

Applied in the solid state formation of an unusual cation deficient perovskite, Ba7Nb4MoO20.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Garcia-Gonzalez, E. et al.
Chemistry of Materials, 11, 433-433 (1999)
Leliz T Arenas et al.
Talanta, 83(1), 241-248 (2010-11-03)
A film of niobium oxide was immobilized on a SiO(2)/C carbon-ceramic matrix (specific surface area 270 m(2)g(-1)) and characterized by N(2) adsorption-desorption isotherms, scanning electron microscopy, X-ray photoelectron spectroscopy and atomic force microscopy. This new carbon-ceramic material, SiO(2)/C/Nb(2)O(5), was used
Ning-Ping Huang et al.
Biomacromolecules, 12(12), 4213-4220 (2011-11-02)
The signal-to-noise ratio of a sensor system is determined by the affinity of its active component for the analyte on one hand and its inertness with respect to unrelated stimuli (noise) on the other hand. Nonspecific interactions between the environment
M Christopher Orilall et al.
Journal of the American Chemical Society, 131(26), 9389-9395 (2009-07-02)
Catalyst-electrode design is crucial for the commercialization and widespread use of polymer electrolyte membrane fuel cells. There are considerable challenges in making less expensive, more durable, and more active catalysts. Herein, we report the one-pot synthesis of Pt and Pt-Pb
Scott B Ficarro et al.
Analytical chemistry, 80(12), 4606-4613 (2008-05-22)
Proteomics-based analysis of signaling cascades relies on a growing suite of affinity resins and methods aimed at efficient enrichment of phosphorylated peptides from complex biological mixtures. Given the heterogeneity of phosphopeptides and the overlap in chemical properties between phospho- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service