Skip to Content
Merck
All Photos(6)

Documents

204390

Sigma-Aldrich

Silver nitrate

99.9999% trace metals basis

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor density

5.8 (vs air)

Quality Level

Assay

99.9999% trace metals basis

form

crystalline
solid

impurities

≤1.5 ppm Trace Metal Analysis

mp

212 °C (dec.) (lit.)

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nitrate is a water-soluble silver salt used as a precursor to prepare silver-containing nanomaterials or complexes.

Application

Silver nitrate can be used as:
  • An activator in Ni-P coating of fly-ash cenosphere particles by the electroless method.
  • A silver precursor to prepare Ag–TiO2 nanomaterials applicable in the photocatalytic degradation of rhodamine 6G dye molecule.
It can also be used as a catalyst in the:
  • Hydrolytic oxidation of organosilanes to generate hydrogen.
  • Oxidation of a variety of aldehydes to corresponding carboxylic acids in the presence of H2O2 as an oxidizer.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Jessl et al.
Small (Weinheim an der Bergstrasse, Germany), 14(20), e1703879-e1703879 (2018-04-18)
Seed-mediated methods employing cetyltrimethylammonium bromide (CTAB) as a surfactant, and silver salts as additives, are the most common synthetic strategies for high-yield productions of quality Au nanorods. However, the mechanism of these reactions is not yet fully understood and, importantly
Andrey Massarsky et al.
Chemosphere, 92(1), 59-66 (2013-04-04)
Nanomaterials (NMs) including silver nanoparticles (AgNPs) are incorporated into an increasing number of consumer and medical products. However, the potential toxicity of AgNPs to aquatic organisms is largely unknown. This study characterizes the effects of AgNPs on zebrafish (Danio rerio)
Konno, T.; et al.
Chemistry Letters (Jpn), 35 (3), 316-317 (2006)
B Vaseeharan et al.
Letters in applied microbiology, 50(4), 352-356 (2010-02-06)
To determine the antibacterial potential of silver nanoparticles (AgNps) synthesized by tea leaf extract against Vibrio harveyi and its protective effect on juvenile Feneropenaeus indicus. AgNps were synthesized by a simple procedure using tea leaf extract as the reducing agent.
Yun Ju Chae et al.
Aquatic toxicology (Amsterdam, Netherlands), 94(4), 320-327 (2009-08-25)
The increased use of nano-sized metallic materials is likely to result in the release of these particles into the environment. It is, however, unclear if these materials are harmful to aquatic animals. Furthermore, because the dissolution of such nanomaterials will

Articles

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

See All

Related Content

This application note discusses the synthesis of silver nanowires.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service