Skip to Content
Merck
All Photos(1)

Key Documents

202533

Sigma-Aldrich

1,3-Butadiene diepoxide

97%

Synonym(s):

1,2,3,4-Diepoxybutane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
Beilstein:
8614599
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

25 mmHg ( 56 °C)

Assay

97%

form

liquid

impurities

<3% methylene chloride

refractive index

n20/D 1.434 (lit.)

bp

56-58 °C/25 mmHg (lit.)

mp

2-4 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1OC1C2CO2

InChI

1S/C4H6O2/c1-3(5-1)4-2-6-4/h3-4H,1-2H2

InChI key

ZFIVKAOQEXOYFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Butadiene diepoxide is a versatile organic compound characterized by its diepoxide structure, which features two epoxy groups. This unique configuration allows it to act as an effective crosslinking agent in polymerization processes, enhancing the mechanical strength and thermal stability of resulting materials.

Application

1,3-Butadiene diepoxide can be used:
  • As a precursor to synthesize microstructured poly(amino alcohol ethers) hydrogels for drug delivery and tissue engineering. It helps to enhance mechanical strength and thermal responsiveness of hydrogels.
  • As a cross-linker for the green synthesis of hemostatic epoxy-amine sponge for management of bleeding. It facilitates the formation of a three-dimensional network within the epoxy-amine matrix.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 1B - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Erin D Michaelson-Richie et al.
Journal of proteome research, 9(9), 4356-4367 (2010-07-30)
1,2,3,4-diepoxybutane (DEB) is a strongly genotoxic diepoxide hypothesized to be the ultimate carcinogenic metabolite of the common industrial chemical and environmental carcinogen 1,3-butadiene. DEB is a bis-electrophile capable of cross-linking cellular biomolecules to form DNA-DNA and DNA-protein cross-links (DPCs), which
Filipa Ponte et al.
Toxicology, 289(1), 52-58 (2011-08-03)
The biotransformation and oxidative stress may contribute to 1,2:3,4-diepoxybutane (DEB)-induced toxicity to human lymphocytes of Fanconi Anemia (FA) patients. Thus, the identification of putative inhibitors of bioactivation, as well as the determination of the protective role of oxidant defenses, on
Qingjun Liu et al.
Biosensors & bioelectronics, 26(4), 1672-1678 (2010-10-15)
Olfactory systems of human beings and animals have the abilities to sense and distinguish varieties of odors. In this study, a bioelectronic nose was constructed by fixing biological tissues onto the surface of light-addressable potentiometric sensor (LAPS) to mimic human
Uthpala Seneviratne et al.
Chemical research in toxicology, 23(10), 1556-1567 (2010-09-30)
1,2,3,4-Diepoxybutane (DEB) is a carcinogenic metabolite of 1,3-butadiene (BD), an important industrial and environmental chemical present in urban air and in cigarette smoke. DEB is considered the ultimate carcinogenic species of BD because of its potent genotoxicity and mutagenicity attributed
Maria Castella et al.
Journal of medical genetics, 48(4), 242-250 (2011-01-11)
Fanconi anaemia (FA) is a rare syndrome characterized by bone marrow failure, malformations and cancer predisposition. Chromosome fragility induced by DNA interstrand crosslink (ICL)-inducing agents such as diepoxybutane (DEB) or mitomycin C (MMC) is the 'gold standard' test for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service