Skip to Content
Merck
All Photos(1)

Documents

199222

Sigma-Aldrich

Chromone

99%

Synonym(s):

1-Benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

55-60 °C (lit.)

SMILES string

O=C1C=COc2ccccc12

InChI

1S/C9H6O2/c10-8-5-6-11-9-4-2-1-3-7(8)9/h1-6H

InChI key

OTAFHZMPRISVEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The compounds with chromone skeleton exhibits antiradical activities that provide protection against oxidative stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra Kładna et al.
Luminescence : the journal of biological and chemical luminescence, 29(7), 846-853 (2014-02-01)
Recent reviews evidence that the naturally occurring compounds containing the chromone skeleton exhibit antiradical activities, providing protection against oxidative stress. The antioxidant activities of 13 new synthesized chromonyl-2,4-thiazolidinediones, chromonyl-2,4-imidazolidinediones and chromonyl-2-thioxoimidzolidine-4-ones were evaluated using in vitro antioxidant assays, including superoxide
Miguel Pinto et al.
Pharmaceutics, 11(7) (2019-07-25)
The current pharmacological treatments for Parkinson's disease only offer symptomatic relief to the patients and are based on the administration of levodopa and catechol-O-methyltransferase or monoamine oxidase-B inhibitors (IMAO-B). Since the majority of drug candidates fail in pre- and clinical
Sarvesh Kumar et al.
Bioorganic & medicinal chemistry, 15(8), 2952-2962 (2007-02-27)
The interaction between leukocytes and the vascular endothelial cells (EC) via cellular adhesion molecules plays an important role in various inflammatory and immune diseases. The molecules that block these interactions have been targeted as potential therapeutic targets for acute and
Anna Chen et al.
Molecules (Basel, Switzerland), 24(17) (2019-08-25)
Isorhamnetin-3-O-rhamnoside was synthesized by a highly efficient three-enzyme (rhamnosyltransferase, glycine max sucrose synthase and uridine diphosphate (UDP)-rhamnose synthase) cascade using a UDP-rhamnose regeneration system. The rhamnosyltransferase gene (78D1) from Arabidopsis thaliana was cloned, expressed, and characterized in Escherichia coli. The
Joana Rocha-Pereira et al.
Bioorganic & medicinal chemistry, 18(12), 4195-4201 (2010-06-18)
Human noroviruses (NoV) are now recognized as the most frequent cause of outbreaks and sporadic cases of acute gastroenteritis. Despite the significant economic impact and considerable morbidity of norovirus disease, no drug or vaccine is currently available to treat or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service