Skip to Content
Merck
All Photos(1)

Key Documents

196320

Sigma-Aldrich

3-Acetylthiophene

98%

Synonym(s):

Methyl-3-thienyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6OS
CAS Number:
Molecular Weight:
126.18
Beilstein:
107241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

208-210 °C/748 mmHg (lit.)

mp

57-62 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1ccsc1

InChI

1S/C6H6OS/c1-5(7)6-2-3-8-4-6/h2-4H,1H3

InChI key

RNIDWJDZNNVFDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Acetylthiophene modified glassy carbon electrode has been used in voltammetric determination of uric acid in urine samples.

Application

3-Acetylthiophene was used in the preparation of 1-(methylthiophenylidine)-8-naphthylamine(Schiff′s base) and 1-(thiophen-3-yl)ethanone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ewa Rozycka-Sokolowska et al.
Acta crystallographica. Section C, Crystal structure communications, 67(Pt 6), o209-o211 (2011-06-03)
The structure of the title compound, C(6)H(6)OS, exhibits a flip-type disorder of the thiophene ring [occupancy ratio = 0.848 (3):0.152 (3)], which is typical for many thiophene derivatives. The puckered thiophene ring is essentially coplanar with the plane formed by
Mehmet Aslanoglu et al.
Chemical & pharmaceutical bulletin, 56(3), 282-286 (2008-03-04)
A reliable and reproducible method for the determination of uric acid in urine samples has been developed. The method is based on the modification of a glassy carbon electrode by 3-acetylthiophene using cyclic voltammetry. The poly(3-acetylthiophene) modified glassy carbon electrode
Electro-oxidative polymerization of Schiff-base of 1, 8-diaminonaphthaline and 3-acetylthiophene. I. Preparation and study the redox behaviour of the resulting polymer.
Hathoot AA.
Eur. Polymer J., 36(5), 1063-1071 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service