Skip to Content
Merck
All Photos(2)

Key Documents

192120

Sigma-Aldrich

Borane dimethyl sulfide complex solution

2.0 M in THF

Synonym(s):

BMS, Trihydro[thiobis[methane]]boron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S · BH3
CAS Number:
Molecular Weight:
75.97
Beilstein:
3663489
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

2.0 M in THF

density

0.855 g/mL at 25 °C

functional group

thioether

storage temp.

2-8°C

SMILES string

B.CSC

InChI

1S/C2H6S.BH3/c1-3-2;/h1-2H3;1H3

InChI key

RMHDLBZYPISZOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Borane dimethyl sulfide complex (BMS) acts as a highly efficient and selective reducing agent in the presence of catalytic sodium tetrahydroborate for α-hydroxy esters. Asymmetric borane reduction of a variety of prochiral ketones with BMS using spiroborate esters as catalyst has been reported.

Application

Borane dimethyl sulfide complex (BMS) solution may be used in the following studies:
  • One-pot conversion of alkynes into 1,2-diols.
  • Preparation of iminopentitols.
  • Reduction of representative organic functional groups, such as esters, nitriles and amides.
  • Used along with a dendrimeric supported L-pyrrolidinol in the asymmetric reduction of indanones and tetralones.
  • Asymmetric reduction of ferrocenyl-1,3-diketones to chiral 1,3-diols.
  • Highly enantioselective reduction of ketones catalyzed by C3-symmetric tripodal hydroxyamides.
Reactant used as a regioselective reducing agent

Reactant involved in:
  • Hydroboration / oxidation
Synthesis of cage compounds

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Viatcheslav Stepanenko et al.
Tetrahedron letters, 48(33), 5799-5802 (2007-08-13)
Novel spiroborate esters derived nonracemic 1,2-aminoalcohols and ethylene glycol are reported as highly effective catalysts for the asymmetric borane reduction of a variety of prochiral ketones with borane-dimethyl sulfide complex at room temperature. Optically active alcohols were obtained in excellent
M Godskesen et al.
Bioorganic & medicinal chemistry, 4(11), 1857-1865 (1996-11-01)
The four stereoisomeric 1,5-dideoxy-1,5-iminopentitols with D-arabino-(D-lyxo-) (3), ribo- (9), L-lyxo (L-arabino-) (13) and xylo-(18) configurations were synthesized. The corresponding aldonolactones (1, 7 and 11) or aldonic acid ester (15b) having a leaving group at C-5 gave by reaction with aqueous
Ana M Gómez et al.
Organic letters, 4(3), 383-386 (2002-02-01)
Primary ozonides derived from alkenyl stannanes display an unusual stability and can be transformed into 1,2-diols by treatment with dimethyl sulfide and borane-methyl sulfide complex. This observation has been incorporated into the development of a novel one-pot strategy for the
Selective reductions. 29. A simple technique to achieve an enhanced rate of reduction of representative organic compounds by borane-dimethyl sulfide.
Brown HC, et al.
The Journal of Organic Chemistry, 47(16), 3153-3163 (1982)
Highly Enantioselective Borane Reduction of Prochiral Ketones Catalyzed by C3-Symmetric Tripodal β-Hydroxy Amides
Fang T, et al.
Synlett, 1559-1559 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service