Skip to Content
Merck
All Photos(3)

Key Documents

191647

Sigma-Aldrich

3,5-Dinitrobenzyl chloride

97%

Synonym(s):

α-Chloro-3,5-dinitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CH2Cl
CAS Number:
Molecular Weight:
216.58
Beilstein:
2506587
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

79-82 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(CCl)cc(c1)[N+]([O-])=O

InChI

1S/C7H5ClN2O4/c8-4-5-1-6(9(11)12)3-7(2-5)10(13)14/h1-3H,4H2

InChI key

SMJODKZAFKWUJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dinitrobenzyl chloride was used in the synthesis of double-spanned double cavity calix[4]arenes and 3,5-dinitrobenzylamines. It also was used in fluorescence flow immunoassay for determination of 4-nitrophenol.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of non-symmetrical 3, 5-diamidobenzyl amines, ethers and sulfides.
Barker D, et al.
Tetrahedron Letters, 49(10), 1660-1664 (2008)
Competitive flow immunoassay with fluorescence detection for determination of 4-nitrophenol.
Nistor C, et al.
Analytica Chimica Acta, 426(2), 185-195 (2001)
Calixarenes. 27. Synthesis, characterization, and complexation studies of double-cavity calix [4] arenes.
Gutsche CD and See KA.
The Journal of Organic Chemistry, 57(16), 4527-4539 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service