Skip to Content
Merck
All Photos(1)

Key Documents

187305

Sigma-Aldrich

4-Methoxyphenethylamine

≥98%

Synonym(s):

2-(4-Methoxyphenyl)ethylamine, 4-Methoxyphenethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
151.21
Beilstein:
508967
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

liquid

refractive index

n20/D 1.538 (lit.)

bp

138-140 °C/20 mmHg (lit.)
254-256 °C

density

1.031 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

COc1ccc(CCN)cc1

InChI

1S/C9H13NO/c1-11-9-4-2-8(3-5-9)6-7-10/h2-5H,6-7,10H2,1H3

InChI key

LTPVSOCPYWDIFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methoxyphenethylamine inhibits the monoamine oxidase-catalyzed deamination of both tyramine and tryptamine.

4-Methoxyphenethylamine is used as a precursor for the synthesis of other organic compounds by the alkylation reaction.

Application

4-Methoxyphenethylamine was used in the synthesis of :
  • pyrrolo[3,2-c]carbazole
  • poly(4-methoxyphenethylamine), required for the immobilization of nitrogenated bases and oligonucleotides
  • organopolyphosphazenes such as poly[bis(4-methoxy benzylamino)polyphosphazene] and poly[bis(4-methoxyphenethylamino)polyphosphazene]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and characterization of novel polyorganophosphazenes substituted with 4-methoxybenzylamine and 4-methoxyphenethylamine for in vitro release of indomethacin and 5-fluorouracil.
Gudasi KB, et al.
Reactive functional Polymers, 66(10), 1149-1157 (2006)
GC--MS analysis of N-(bromodimethoxybenzyl)-2-, 3-, and 4-methoxyphenethylamines: Inverse analogues of the psychoactive 25B-NBOMe drug
Almalki AJ, et al.
Forensic Chemistry, 21, 100277-100277 (2020)
Francielle B Silva et al.
International journal of molecular sciences, 9(7), 1173-1188 (2009-03-28)
This work describes the immobilization of purine and pyrimidine bases and immobilization/hybridization of synthetic oligonucleotides on graphite electrodes modified with poly(4-methoxyphenethylamine) produced in acid medium. The immobilization of adenine, guanine, cytosine and thymine on these modified electrodes was efficient, producing
F Peter Guengerich et al.
The Journal of biological chemistry, 277(37), 33711-33719 (2002-07-03)
Cytochrome P450 (P450) 2D6 is involved in the oxidation of a large fraction ( approximately 30%) of drugs used by humans and also catalyzes the O-demethylation of the model substrates 3- and 4-methoxyphenethylamine followed by subsequent ring hydroxylation to dopamine.
T Kuramoto et al.
Journal of neuroscience research, 6(1), 37-48 (1981-01-01)
K+-sensitive liquid ion exchange electrode systems respond with a slow potential change to acetylcholine, choline, anticholinergic drugs, biogenic amines, and glutamic acid. The response threshold has been defined, and in most cases it is at extremely low concentrations (10(-7)-10(-5)M). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service