Skip to Content
Merck
All Photos(1)

Key Documents

183067

Sigma-Aldrich

6-Methoxyquinoline

98%

Synonym(s):

6-Methoxyquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO
CAS Number:
Molecular Weight:
159.18
Beilstein:
115244
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.625 (lit.)

bp

140-146 °C/15 mmHg (lit.)

mp

18-20 °C (lit.)

density

1.15 g/mL at 20 °C (lit.)

SMILES string

COc1ccc2ncccc2c1

InChI

1S/C10H9NO/c1-12-9-4-5-10-8(7-9)3-2-6-11-10/h2-7H,1H3

InChI key

HFDLDPJYCIEXJP-UHFFFAOYSA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

Application

6-Methoxyquinoline is used as a precursor in the synthesis of:
  • Fluorescent zinc and chlorine sensors.
  • 5-Amino-2-aroylquinolines as potent tubulin polymerization inhibitors.
  • 3-Fluoro-6-methoxyquinoline derivatives as inhibitors of bacterial DNA gyrase and topoisomerase.
  • Cobalt-based ternary metal-organic complex as single?ion magnets.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Novel 3-fluoro-6-methoxyquinoline derivatives as inhibitors of bacterial DNA gyrase and topoisomerase IV.
Mitton-Fry MJ, et al.
Bioorganic & Medicinal Chemistry Letters, 27(15), 3353-3358 (2017)
Roland J Reischl et al.
Journal of chromatography. A, 1269, 262-269 (2012-08-28)
The determination of trace amounts of d-amino acids (d-AAs) even in tissue samples of higher developed animals, mammals and humans has opened a wide field of biological questions to be investigated. d-Ala, d-Asp and d-Ser have already been identified to
Single-Ion Magnets Based on Mononuclear Cobalt (II) Complexes with Sulfadiazine.
Villa-Perez C, et al.
European Journal of Inorganic Chemistry, 2016(29), 4835-4841 (2016)
Richard G Painter et al.
Analytical chemistry, 78(9), 3133-3137 (2006-04-29)
Human neutrophils synthesize hypochlorous acid, a nonradical oxidant, as one of the antimicrobial agents to kill phagocytosed pathogens within phagolysosomes. The production of HOCl is catalyzed by myeloperoxidase using chloride anions (Cl-). Even though various approaches have been documented to
Methoxy-substituted TQEN family of fluorescent zinc sensors.
Mikata Y, et al.
Inorganic Chemistry, 45(23), 9262-9268 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service