Skip to Content
Merck
All Photos(2)

Key Documents

177490

Sigma-Aldrich

16-Hydroxyhexadecanoic acid

98%

Synonym(s):

16-Hydroxypalmitic acid, Juniperic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)15CO2H
CAS Number:
Molecular Weight:
272.42
Beilstein:
1783998
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

94-98 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

OCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O3/c17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16(18)19/h17H,1-15H2,(H,18,19)

InChI key

UGAGPNKCDRTDHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

16-Hydroxyhexadecanoic acid was used in the synthesis of dihydroxypalmitic acids. It was also used to induce the expression of two GRP genes of Arabidopsis thaliana, AtGRP5 and AtGRP23.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Grausem et al.
Plant, cell & environment, 37(9), 2102-2115 (2014-02-14)
Cutin and suberin represent lipophilic polymers forming plant/environment interfaces in leaves and roots. Despite recent progress in Arabidopsis, there is still a lack on information concerning cutin and suberin synthesis, especially in crops. Based on sequence homology, we isolated two
Peiyao Zhu et al.
Journal of cell communication and signaling, 14(2), 175-192 (2020-01-12)
Esophageal squamous cell carcinoma (ESCC) is one of the most common malignant tumors with poor prognosis. Aryl hydrocarbon receptor (AHR) is a ligand-dependent transcription factor and emerging evidence shows it is associated with tumor initiation and promotion. However, the relationship
Nejumal K Khalid et al.
Environmental monitoring and assessment, 190(6), 370-370 (2018-06-02)
The presence of emerging contaminants (ECs) in different aquatic systems may contribute to hazardous effects on aquatic organisms and subsequently on human health. In the present work, liquid chromatography coupled to a quadrupole time of flight mass spectrometer (LC-Q-ToF-MS) was
The importance of water to biocatalysis in organic solvents.
M J Alston et al.
Biochemical Society transactions, 23(1), 70S-70S (1995-02-01)
Jong Ho Park et al.
Plant physiology and biochemistry : PPB, 46(11), 1015-1018 (2008-07-29)
Glycine-rich proteins (GRPs) belong to a large family of heterogenous proteins that are enriched in glycine residues. The expression of two GRP genes of Arabidopsis thaliana, AtGRP5 and AtGRP23, was induced by 16-hydroxypalmitic acid (HPA), a major component of cutin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service