All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C15H10O
CAS Number:
Molecular Weight:
206.24
Beilstein:
608049
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
118-122 °C (lit.)
functional group
ketone
phenyl
SMILES string
O=C1C(c2ccccc2)=C1c3ccccc3
InChI
1S/C15H10O/c16-15-13(11-7-3-1-4-8-11)14(15)12-9-5-2-6-10-12/h1-10H
InChI key
HCIBTBXNLVOFER-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Diphenylcyclopropenone is a potent hapten. Its vibration frequency has been evaluated.
Application
Diphenylcyclopropenone was used in topical immunotherapy.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Valerio Tabacchini et al.
Physics in medicine and biology, 60(14), 5513-5525 (2015-07-03)
Gamma-ray detectors based on bright monolithic scintillation crystals coupled to pixelated photodetectors are currently being considered for several applications in the medical imaging field. In a typical monolithic detector, both the light intensity and the time of arrival of the
Release of erythropoietin and neuron-specific enolase after breath holding in competing free divers.
T Kjeld et al.
Scandinavian journal of medicine & science in sports, 25(3), e253-e257 (2014-08-22)
Free diving is associated with extreme hypoxia. This study evaluated the combined effect of maximal static breath holding and underwater swimming on plasma biomarkers of tissue hypoxemia: erythropoietin, neuron-specific enolase and S100B, C-reactive protein, pro-atrial natriuretic peptide, and troponin T.
Yunseok Choi et al.
Annals of dermatology, 25(4), 434-439 (2013-12-29)
There exists a treatment challenge with periungual warts. Topical immunotherapy with diphenylcyclopropenone (DPCP) has recently been reported to be an effective treatment for recalcitrant warts, including periungual types. We aimed to evaluate the effectiveness and preference of topical immunotherapy with
Shigeki Inui et al.
The Journal of dermatology, 37(10), 888-893 (2010-09-24)
Pigmentary complication by contact immunotherapy (CI) for alopecia areata (AA) has been reported but its pathophysiology remains unknown. To characterize pigmentary complication by CI and its pathophysiology, we examined the incidence of hyperpigmentation in 186 consecutive patients treated with CI
Nannie Bangsgaard et al.
Contact dermatitis, 63(1), 10-14 (2010-07-06)
It is uncertain whether polysensitized patients acquire multiple allergies only because of a high degree of exposure to environmental allergens, or because of being highly susceptible to developing contact allergy. The aim of this study was to investigate and compare
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service