Skip to Content
Merck
All Photos(1)

Key Documents

176761

Sigma-Aldrich

Potassium deuteroxide solution

40 wt. % in D2O, 98 atom % D

Synonym(s):

Potassium hydroxide-d solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOD
CAS Number:
Molecular Weight:
57.11
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

form

liquid

concentration

40 wt. % in D2O

technique(s)

NMR: suitable

refractive index

n20/D 1.411

mass shift

M+1

SMILES string

[K+].[O-][2H]

InChI

1S/K.H2O/h;1H2/q+1;/p-1/i/hD

InChI key

KWYUFKZDYYNOTN-DYCDLGHISA-M

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Fortes
Physical chemistry chemical physics : PCCP, 21(16), 8264-8274 (2019-04-04)
Water ice is one of the most fundamental and well-studied molecular solids, which continues to provide surprising and useful insights into its emergent complexity as ever more precise experimental techniques are applied. Using one of the highest resolution neutron powder

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service