Skip to Content
Merck
All Photos(1)

Key Documents

17356

Sigma-Aldrich

3-(Boc-amino)propyl bromide

≥96.0% (GC)

Synonym(s):

tert-Butyl N-(3-bromopropyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)3NHCO2C(CH3)3
CAS Number:
Molecular Weight:
238.12
Beilstein:
4176344
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (GC)

reaction suitability

reagent type: cross-linking reagent

mp

37-39 °C

functional group

Boc
amine
bromo

storage temp.

2-8°C

SMILES string

BrCCCNC(OC(C)(C)C)=O

InChI

1S/C8H16BrNO2/c1-8(2,3)12-7(11)10-6-4-5-9/h4-6H2,1-3H3,(H,10,11)

InChI key

IOKGWQZQCNXXLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Boc-amino)propyl bromide can be used as an alkylating reagent for the synthesis of:
  • Benzydamine analogs to be used as activators for soluble guanylate cyclase.
  • N-substituted chromenotriazolopyrimidine, human murine double minute 2 (MDM2) inhibitor.
  • Protected amines from piperidine derivatives to be further used for synthesis of sulfonamide series.

It can also be used for the post-polymerization quaternization of polymers to synthesize functional cationic polymers and antimicrobial agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of novel pyrazoles and indazoles as activators of the nitric oxide receptor, soluble guanylate cyclase.
Selwood DL, et al.
Journal of medicinal chemistry, 44(1), 78-93 (2001)
Identification of selective 8-(piperidin-4-yloxy) quinoline sulfone and sulfonamide histamine H1 receptor antagonists for use in allergic rhinitis.
Procopiou PA, et al.
Bioorganic & Medicinal Chemistry Letters, 27(21), 4914-4919 (2017)
Improvement of the synthesis and pharmacokinetic properties of chromenotriazolopyrimidine MDM2-p53 protein-protein inhibitors.
Beck HP, et al.
Bioorganic & Medicinal Chemistry Letters, 21(9), 2752-2755 (2011)
Design, syntheses and evaluation of hemocompatible pegylated-antimicrobial polymers with well-controlled molecular structures.
Venkataraman S, et al.
Biomaterials, 31(7), 1751-1756 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service