Skip to Content
Merck
All Photos(2)

Key Documents

160989

Sigma-Aldrich

3-Ethoxysalicylaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H3-2-(OH)CHO
CAS Number:
Molecular Weight:
166.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

263-264 °C (lit.)

mp

66-68 °C (lit.)

SMILES string

CCOc1cccc(C=O)c1O

InChI

1S/C9H10O3/c1-2-12-8-5-3-4-7(6-10)9(8)11/h3-6,11H,2H2,1H3

InChI key

OFQBYHLLIJGMNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Ethoxysalicylaldehyde was used in the preparation of:
  • Schiff base ligand via condensation with trans-1,2-diaminocyclohexane
  • substituted salen-type Schiff base ligands
  • a new Schiff base, 2-ethoxy-6-[(3-methylaminopropylimino)methyl]phenol, via reaction with N-methylpropane-1,3-diamine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melita Lončarić et al.
Current organic synthesis, 17(2), 98-108 (2020-05-19)
In order to preserve the environment from harmful organic solvents, a synthesis of coumarin derivatives was performed in deep eutectic solvents, which are considered as "green" due to their characteristics. Choline chloride based deep eutectic solvents (DESs) were employed, both
Arpita Jana et al.
Inorganic chemistry, 49(19), 9012-9025 (2010-09-04)
Syntheses, characterization, and magnetic properties of a series of diphenoxo-bridged discrete dinuclear M(II)Ln(III) complexes (M = Cu or Ni, Ln = Ce-Yb) derived from the compartmental Schiff base ligand, H(2)L, obtained on condensation of 3-ethoxysalicylaldehyde with trans-1,2-diaminocyclohexane, are described. Single
Hydroxylation of phenol catalyzed by oxovanadium (IV) of salen-type schiff base complexes with hydrogen peroxide.
Alsalim TA, et al.
Catalysis Letters, 136(3-4), 228-233 (2010)
Synthesis, Crystal Structures, and Antibacterial Activities of Schiff-Base Zinc (II) Complexes [ZnL1Cl2] and [ZnL2I2]? 0.5 CH3OH.
Guo Y-N.
Synthesis and Reactivity in Inorganic, Metal-Organic, and Nano-Metal Chemistry, 41(8), 987-991 (2011)
Tuhin Khan et al.
Physical chemistry chemical physics : PCCP, 19(44), 30120-30127 (2017-11-04)
Salophen (N,N'-bis-(salicylidene)-o-phenylenediamine) is a Schiff base, which is weakly emissive in solution, but strongly emissive in the solid state. The trend is reversed in its aluminium complex (SalAl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service