15879
Octaethylene glycol
≥95% (oligomer purity)
Synonym(s):
PED-diol (n=8)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO(CH2CH2O)8H
CAS Number:
Molecular Weight:
370.44
EC Number:
MDL number:
UNSPSC Code:
12161502
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95% (oligomer purity)
reaction suitability
reagent type: cross-linking reagent
SMILES string
OCCOCCOCCOCCOCCOCCOCCOCCO
InChI
1S/C16H34O9/c17-1-3-19-5-7-21-9-11-23-13-15-25-16-14-24-12-10-22-8-6-20-4-2-18/h17-18H,1-16H2
InChI key
GLZWNFNQMJAZGY-UHFFFAOYSA-N
General description
Octaethylene glycol also known as PED-diol (n=8), is a cross linking reagent commonly used in organic synthesis.
Application
Octaethylene glycol is used as a cross-linking reagent to produce cyclic octaethylene glycol 5-nitroisophthalate. It serves as a starting material for synthesizing heterobifunctionalized clickable oligo(ethylene glycol) linkers and supramolecular polymers.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Monomers for non-bond crosslinking of vinyl polymers: II. Cyclic octaethylene glycol 5-methacrylamido-isophthalate
A Zada
European Polymer Journal, 36, 351-357 (2000)
Efficient synthesis of diverse heterobifunctionalized clickable oligo (ethylene glycol) linkers: potential applications in bioconjugation and targeted drug delivery
LN Goswami
Organic & Biomolecular Chemistry, 11, 1116-1126 (2013)
Tamiru Negash Alkie et al.
PloS one, 13(11), e0206324-e0206324 (2018-11-02)
Mucosal vaccine delivery systems have paramount importance for the induction of mucosal antibody responses. Two studies were conducted to evaluate immunogenicity of inactivated AIV antigens encapsulated in poly(D,L-lactide-co-glycolide) (PLGA) nanoparticles (NPs). In the first study, seven groups of specific pathogen
Pedro Cecílio et al.
PLoS neglected tropical diseases, 11(11), e0005951-e0005951 (2017-11-28)
The notion that previous infection by Leishmania spp. in endemic areas leads to robust anti-Leishmania immunity, supports vaccination as a potentially effective approach to prevent disease development. Nevertheless, to date there is no vaccine available for human leishmaniasis. We optimized
An azobenzene-based single-component supramolecular polymer responsive to multiple stimuli in water
E Fuentes
Journal of the American Chemical Society, 142, 10069-10078 (2020)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service