Skip to Content
Merck
All Photos(2)

Documents

157724

Sigma-Aldrich

2-Ethyl-6-methylaniline

≥98%

Synonym(s):

3-Ethyl-2-aminotoluene, 6-Ethyl-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H3(CH3)NH2
CAS Number:
Molecular Weight:
135.21
Beilstein:
2079468
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.06 mmHg ( 20 °C)

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.552 (lit.)

bp

231 °C (lit.)

mp

−33 °C (lit.)

density

0.968 g/mL at 25 °C (lit.)

SMILES string

CCc1cccc(C)c1N

InChI

1S/C9H13N/c1-3-8-6-4-5-7(2)9(8)10/h4-6H,3,10H2,1-2H3

InChI key

JJVKJJNCIILLRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethyl-6-methylaniline is metabolite of herbicides metolachlor, diuron, monuron and its degradation was assessed in laboratory experiments on microbiologically active and sterilized soils.

Application

2-Ethyl-6-methylaniline was used in the preparation of racemic esters of N-(2-ethyl-6-methylphenyl)alanine (NEMPA).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

221.0 °F

Flash Point(C)

105 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resolution of< i> N</i>-(2-ethyl-6-methylphenyl) alanine via free and immobilized lipase from< i> Pseudomonas cepacia</i>.
Zheng L, et al.
Journal of Molecular Catalysis. B, Enzymatic, 38(3), 119-125 (2006)
A Barra Caracciolo et al.
Environmental pollution (Barking, Essex : 1987), 134(3), 525-534 (2004-12-29)
In this work the degradation of the herbicides metolachlor, diuron, monuron and of the metabolites 2-ethyl-6-methylaniline (EMA), and 3,4-dichloroaniline (DCA) was assessed in laboratory experiments on microbiologically active and sterilized soils. Their leaching potentials were calculated, using Gustafson's equation, by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service