Skip to Content
Merck
All Photos(1)

Key Documents

157384

Sigma-Aldrich

Phenanthridine

98%

Synonym(s):

Benzo[c]quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9N
CAS Number:
Molecular Weight:
179.22
Beilstein:
120204
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

bp

349 °C/769 mmHg (lit.)

mp

104-107 °C (lit.)

SMILES string

c1ccc2c(c1)cnc3ccccc23

InChI

1S/C13H9N/c1-2-6-11-10(5-1)9-14-13-8-4-3-7-12(11)13/h1-9H

InChI key

RDOWQLZANAYVLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Used in the study of ecotoxicity.
  • Used in rotational spectroscopic investigations and radio astronomical searches.
  • Used as a dye in biological and chemical sensors.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural-based differences in ecotoxicity of benzoquinoline isomers to the zebra mussel (Dreissena polymorpha).
Kraak, Michiel HS, et al.
Environmental Toxicology and Chemistry / Setac, 16(10), 2158-2163 (1997)
Jian Zhang et al.
The journal of physical chemistry. B, 109(18), 8701-8706 (2006-07-21)
Tiopronin-coated silver particles (average diameters of core = 1.6 nm) were prepared by a modified Brust method, and the ligands on the metal core were partially displaced by (2-mercapto-propionylamino) acetic acid-2,5-dioxo-pyrrolidin-1-ylester through ligand exchanges. The particles were bound on amine-pendent
FT-MW and Millimeter Wave Spectroscopy of PANHs: Phenanthridine, Acridine, and 1, 10-Phenanthroline
McNaughton, Don, et al.
The Astrophysical Journal, 678(1), 309-309 (2008)
Non-aggregating solvatochromic bipolar benzo [f] quinolines and benzo [a] acridines for organic electronics.
Goel, Atul, et al.
Journal of Materials Chemistry, 22(30), 14880-14888 (2012)
Harrie J M Gijsen et al.
Journal of medicinal chemistry, 53(19), 7011-7020 (2010-09-03)
The TRPA1 channel can be considered as a key biological sensor to irritant chemicals. In this paper, the discovery of 11H-dibenz[b,e]azepines (morphanthridines) and dibenz[b,f][1,4]oxazepines is described as extremely potent agonists of the TRPA1 receptor. This has led to the discovery

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service